Skip to Content
Merck
All Photos(1)

Key Documents

15406

Sigma-Aldrich

N-Boc-cadaverine

≥97.0% (NT)

Synonym(s):

N-Boc-1,5-diaminopentane, tert-Butyl N-(5-aminopentyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH)3COCONH(CH2)5NH2
CAS Number:
Molecular Weight:
202.29
Beilstein:
3603658
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (NT)

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.460

density

0.972 g/mL at 20 °C (lit.)

functional group

Boc
amine

SMILES string

NCCCCCNC(OC(C)(C)C)=O

InChI

1S/C10H22N2O2/c1-10(2,3)14-9(13)12-8-6-4-5-7-11/h4-8,11H2,1-3H3,(H,12,13)

InChI key

DPLOGSUBQDREOU-UHFFFAOYSA-N

Application

Some of the reported applications of N-Boc-cadaverine include:
  • Synthesis of of a supermacrocycle that self-assemble to form organic nanotubes.
  • Preparation of water-soluble unsymmetrical sulforhodamine fluorophores from monobrominated sulfoxanthene dye.
  • Synthesis of functionalized porphyrins as biocompatible carrier system for photodynamic therapy (PDT).

Other Notes

Building block for preparing polyamines and polyamides

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Melchiorre et al.
Journal of medicinal chemistry, 32(1), 79-84 (1989-01-01)
Several polymethylene tetraamines related to methoctramine (1) were prepared and evaluated for their blocking activity on M-2 muscarinic receptors in guinea pig atria and ileum. It turned out that antimuscarinic potency depends on the following parameters: (a) nature of the
V.J. Jasys et al.
The Journal of Organic Chemistry, 57, 1814-1814 (1992)
T. Teshima et al.
Tetrahedron, 47, 3305-3305 (1991)
Helical rosette nanotubes: design, self-assembly, and characterization.
Fenniri H, et al.
Journal of the American Chemical Society, 123(16), 3854-3855 (2001)
A toolset of functionalized porphyrins with different linker strategies for application in bioconjugation.
Staegemann M H, et al.
Organic & Biomolecular Chemistry, 14(38), 9114-9132 (2016)

Articles

Mono-Boc-protected diamines are versatile building blocks for chemical synthesis. Their production is a lot more challenging than the simple reaction scheme might imply, because the Boc-anhydride reagent cannot differentiate between the two identical amino moieties in the substrate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service