Skip to Content
Merck
All Photos(3)

Key Documents

153958

Sigma-Aldrich

3-Bromophenylhydrazine hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4NHNH2·HCl
CAS Number:
Molecular Weight:
223.50
Beilstein:
3565829
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Quality Level

Assay

98%

mp

227-231 °C (dec.) (lit.)

functional group

bromo
hydrazine

SMILES string

Cl.NNc1cccc(Br)c1

InChI

1S/C6H7BrN2.ClH/c7-5-2-1-3-6(4-5)9-8;/h1-4,9H,8H2;1H

InChI key

RPYIPFXHIKXRKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Bromophenylhydrazine hydrochloride was used to prepare 6-(3-Bromophenylhydrazono)-7,8,9,11-tetrahydropyrido [2,1- b]quinazoline-11-one.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and COX inhibitory activities of rutaecarpine derivatives.
Bull. Korean Chem. Soc., 26(12), 1975-1975 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service