152765
9-Anthracenecarbonitrile
97%
Synonym(s):
9-Cyanoanthracene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H9N
CAS Number:
Molecular Weight:
203.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
powder
mp
173-177 °C (lit.)
functional group
nitrile
SMILES string
N#Cc1c2ccccc2cc3ccccc13
InChI
1S/C15H9N/c16-10-15-13-7-3-1-5-11(13)9-12-6-2-4-8-14(12)15/h1-9H
InChI key
KEQZHLAEKAVZLY-UHFFFAOYSA-N
General description
The fluorescence excitation spectra of 9-anthracenecarbonitrile has been studied.
Application
9-Anthracenecarbonitrile was used to study the mechanism of charge separation within phenothiazine (PTZH) or phenoxazine (PXZH), and 9-cyanoanthracene(electron acceptor).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
O A Andreev et al.
Journal of muscle research and cell motility, 16(4), 353-367 (1995-08-01)
A serine residue located in the active site of myosin head (S1) was labelled by 9-anthroylnitrile, an amino group located in the central domain of S1 was labelled by 7-diethylamino-3-(4'-isothio-cyanato-phenyl)-4-methylcoumari n, a cysteine residue located near the C-terminus of S1
K Szarka et al.
Biochemistry, 40(49), 14806-14811 (2001-12-26)
It has been shown that one of the 12 serine residues within the 23 kDa segment of myosin subfragment 1 can be covalently modified with a fluorescent probe 9-anthroylnitrile (ANN) [Hiratsuka, T. (1989) J. Biol. Chem. 264 (30), 18188-18194]. To
Reika Kanya et al.
The Journal of chemical physics, 121(19), 9489-9497 (2004-11-13)
Fluorescence excitation spectra of the S(1)-S(0) origin band of 9-cyanoanthracene have been observed under a uniform electric field up to 200 kV/cm to explore pendular-state spectrum of an asymmetric-top molecule close to the strong field limit. The observed spectra exhibit
M Takahashi et al.
Journal of pharmaceutical and biomedical analysis, 14(11), 1579-1584 (1996-08-01)
9-Anthroylnitrile was used as an achiral reagent for the derivatization of carnitine. The reagent forms UV-absorbing derivatives with the hydroxyl groups of carnitine enantiomers under very mild conditions. The derivatives were separated by high-performance liquid chromatography on an ovomucoid-conjugated column
Barbara Pliszka et al.
Biochimica et biophysica acta, 1699(1-2), 201-205 (2004-05-26)
The fluorescent probe, 9-anthroylnitrile (ANN), can selectively attach to Ser-180 at the ATP-binding site of subfragment 1 (S1) of skeletal muscle myosin [J. Biol. Chem. 278 (2003) 31891]. We have found that MgATP, MgATPgammaS, MgADP.AlF(4) or MgPP(i), but not MgADP
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service