Skip to Content
Merck
All Photos(2)

Documents

15224

Sigma-Aldrich

Bis(trifluoromethane)sulfonimide lithium salt

puriss., ≥99.0% (19F-NMR)

Synonym(s):

LiTFSI, Bis(trifluoromethylsulfonyl)amine lithium salt, Lithium bistrifluoromethanesulfonimidate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO2NLiSO2CF3
CAS Number:
Molecular Weight:
287.09
Beilstein:
6625414
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

puriss.

Assay

≥99.0% (19F-NMR)

form

powder

impurities

≤1% water

mp

234-238 °C (lit.)

solubility

H2O: 10 mg/mL, clear, colorless

SMILES string

[Li]N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F

InChI

1S/C2F6NO4S2.Li/c3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;/q-1;+1

InChI key

QSZMZKBZAYQGRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bis(trifluoromethane)sulfonimide lithium salt (LiTFSI) is a source of lithium (Li+) ions that is extensively used as an organic electrolyte in lithium-ion batteries and capacitors. LiTFSI is also used as an additive to enhance the hole conductivity in perovskite and dye-sensitized solar cells.

Other Notes

Electrolyte suitable for lithium batteries showing very high conductivity

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Nervous system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ionic conductivity in relation to ternary phase diagram of poly (ethylene oxide), succinonitrile, and lithium bis (trifluoromethane) sulfonimide blends.
Echeverri M, et al.
Macromolecules, 45(15), 6068-6077 (2012)
Development of polyacrylonitrile-based polymer electrolytes incorporated with lithium bis (trifluoromethane) sulfonimide for application in electrochromic device.
Sim LN, et al.
Electrochimica Acta, 229(15), 22-30 (2017)
Corina Paraschiva Ciobanu et al.
3 Biotech, 10(7), 298-298 (2020-06-19)
The addition of n-dodecane (between 1-3%) to the Escherichia coli fermentation broth in a mechanically agitated and aerated bioreactor revealed improved DO (dissolved oxygen) levels induced during fermentation which lead to an increase in biomass productivity and faster glucose consumption.
M. Liu et al.
Journal of the Electrochemical Society, 138, 1891-1891 (1991)
A. Webber
Journal of the Electrochemical Society, 138, 2586-2586 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service