Skip to Content
Merck
All Photos(1)

Documents

151971

Sigma-Aldrich

Octane-d18

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)6CD3
CAS Number:
Molecular Weight:
132.34
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

liquid

expl. lim.

0.34-6.3 % (lit.)

technique(s)

NMR: suitable

refractive index

n20/D 1.394 (lit.)

bp

125-127 °C

mp

-57 °C (lit.)

density

0.815 g/mL at 25 °C (lit.)

mass shift

M+18

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C8H18/c1-3-5-7-8-6-4-2/h3-8H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8D2

InChI key

TVMXDCGIABBOFY-VAZJTQEUSA-N

General description

Octane-d18 is a deuterated NMR solvent. It is useful in NMR-based research and analyses. It can be prepared starting from n-octane. It undergoes H/D (Hydrogen/Deuterium) exchange reaction with mono pinacolboryl complex at 110°C.

Application

Octane-d18 has been used to evaluate the separation factors of the deuterated octane isomers by gas chromatography (GC).

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic, thermal, regioselective functionalization of alkanes and arenes with borane reagents.
Hartwig JF.
ACS Symp. Ser., 136-154 (2004)
Gregory P Horne et al.
Dalton transactions (Cambridge, England : 2003), 48(30), 11547-11555 (2019-07-12)
We report a 31P NMR investigation into the activated radioprotection mechanism of octylphenyl-N,N-diisobutyl-carbamoylmethyl phosphine oxide (CMPO) and analogues in the presence of a gamma radiation field. CMPO exhibits significantly enhanced radiation resistance in the presence of high nitric acid concentrations
Deuterium Tracer Studies and Gas Chromatography-Mass Spectrometry Analysis of Deuterated Products during Fischer-Tropsch Synthesis.
Shi B, et al.
Topics in Catalysis, 57(6-9), 460-469 (2014)
The Synthesis and Properties of n-Octane-d18 and 2, 2, 4-Trimethylpentane-d181.
Dixon JA and Marr JW.
The Journal of Organic Chemistry, 28(11), 3085-3087 (1963)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service