Skip to Content
Merck
All Photos(1)

Key Documents

151645

Sigma-Aldrich

Thiazole

99%

Synonym(s):

3-Azathiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3NS
CAS Number:
Molecular Weight:
85.13
Beilstein:
103852
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.538 (lit.)

bp

117-118 °C (lit.)

density

1.2 g/mL at 25 °C (lit.)

SMILES string

c1cscn1

InChI

1S/C3H3NS/c1-2-5-3-4-1/h1-3H

InChI key

FZWLAAWBMGSTSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thiazole is a naturally occurring compound in sesame seed oil and chicken.

Application

Thiazoles were used to create novel recognition motifs for interaction with divalent and trivalent metal ions in siderophores or antibiotics.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thiazole and oxazole peptides: biosynthesis and molecular machinery.
R S Roy et al.
Natural product reports, 16(2), 249-263 (1999-05-20)
Chem. Abstr., 113, 210350s-210350s (1990)
Yukiko Kiniwa et al.
PloS one, 10(5), e0124094-e0124094 (2015-05-21)
Immunotherapy has emerged as a promising strategy for the treatment of metastatic melanoma. Clinical studies have demonstrated the feasibility of cancer immunotherapy using tumor antigens recognized by CD8(+) T cells. However, the overall immune responses induced by these antigens are
Chem. Abstr., 105, 189645q-189645q (1986)
Lebensm. Wiss. Technol., 19, 122-122 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service