Skip to Content
Merck
All Photos(2)

Documents

148997

Sigma-Aldrich

Methyl valerate

99%

Synonym(s):

Methyl pentanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CO2CH3
CAS Number:
Molecular Weight:
116.16
Beilstein:
1741905
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.397 (lit.)

bp

128 °C (lit.)

density

0.875 g/mL at 25 °C (lit.)

SMILES string

CCCCC(=O)OC

InChI

1S/C6H12O2/c1-3-4-5-6(7)8-2/h3-5H2,1-2H3

InChI key

HNBDRPTVWVGKBR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl valerate is a methyl ester of valeric acid, generally used as a fragrance in the manufacture of soaps and laundry detergents. It is also used as a plasticizer and surrogate component for biodiesel.

Application

Methyl valerate can be used:
  • In the preparation of lankacidin C 8-valerate.
  • As a reactant to synthesize bis(2,2,2-trifluoroethyl) 2-oxoalkylphosphonates.
  • As a substrate in the fluorination reactions of unactivated C(sp3)H bonds using decatungstate photocatalyst and N-fluorobenzenesulfonimide.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

84.2 °F

Flash Point(C)

29 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K Nakahama et al.
The Journal of antibiotics, 28(5), 390-394 (1975-05-01)
Lankacidin C, a component of lankacidin-group (T-2636) antibiotics, was esterified to lankacidin C 8-butyrate in the presence of methyl butyrate by culture broth and by cell-free extract of Bacillus megaterium IFO 12108. In addition, methyl isobutyrate, methyl valerate and methyl
Studies on lankacidin-group (T-2636) antibiotics. X
Nakahama K, et al.
The Journal of Antibiotics, 28(5), 390-394 (1975)
A practical process for the preparation of bis (2, 2, 2-trifluoroethyl) 2-oxoalkylphosphonates by acylation with carboxylic esters
Molnar K, et al.
Tetrahedron Letters, 56(34), 4877-4879 (2015)
A Convenient Photocatalytic Fluorination of Unactivated C-H Bonds
Halperin SD, et al.
Angewandte Chemie (International Edition in English), 126(18), 4778-4781 (2014)
Design and control of reactive distillation process for the production of methyl valerate
Chen C-L, et al.
Industrial & Engineering Chemistry Research, 55(5), 1347-1360 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service