Skip to Content
Merck
All Photos(3)

Key Documents

147834

Sigma-Aldrich

Michler′s ketone

98%

Synonym(s):

4,4′-Bis(dimethylamino)benzophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2NC6H4]2CO
CAS Number:
Molecular Weight:
268.35
Beilstein:
790733
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

powder

mp

174-176 °C (lit.)

SMILES string

CN(C)c1ccc(cc1)C(=O)c2ccc(cc2)N(C)C

InChI

1S/C17H20N2O/c1-18(2)15-9-5-13(6-10-15)17(20)14-7-11-16(12-8-14)19(3)4/h5-12H,1-4H3

InChI key

VVBLNCFGVYUYGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Michler′s ketone (MK) is a derivative of benzophenone that shows temperature dependent phosphorescence. It also exhibits triplet-triplet absorption spectra at a lower temperature.

Application

MK can be used as an additive that acts as a photoinitiator in the preparation of dyes. It can also be used as a precursor material in the synthesis of 4,4′-bis{N,N,dimethyl, N (2-ethoxy carbonyl-1-propenyl) ammonium hexafluoro antimonate}benzophenone (MKEA).

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Dam. 1 - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

New Push-Pull Dyes Derived from Michler?s Ketone For Polymerization Reactions Upon Visible Lights.
Tehfe M, et al.
Macromolecules, 46(10), 3761-3770 (2013)
Michler's ketone.
Report on carcinogens : carcinogen profiles, 12, 270-271 (2011-08-24)
Excited state dynamics of Michler's ketone: a laser flash photolysis study
Singh AK, et al.
Research on Chemical Intermediates, 27(1-2), 125-136 (2001)
L G Rushing et al.
Journal of chromatographic science, 18(5), 224-232 (1980-05-01)
Sensitive and specific procedures are described for the analysis of residues of gentian violet in animal feed, human urine, and wastewater at levels of 1000 ppm down to 10 ppb, 1 ppb, and 10 ppb, respectively. The cleaned-up extracts were
Cinzia Chiappe et al.
The journal of physical chemistry. A, 110(14), 4937-4941 (2006-04-08)
Michler's ketone (MK) and tetracyanoethene (TCNE) may be used as a UV-vis probe to investigate the solvent properties of ionic liquids (ILs). In molecular solvents, MK and TCNE give an electron donor-acceptor (EDA) complex, a zwitterionic species or a radical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service