Skip to Content
Merck
All Photos(1)

Documents

146692

Sigma-Aldrich

2-Indanone

98%

Synonym(s):

β-Hydrindone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8O
CAS Number:
Molecular Weight:
132.16
Beilstein:
636550
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

impurities

≤2.0% water

mp

51-54 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C1Cc2ccccc2C1

InChI

1S/C9H8O/c10-9-5-7-3-1-2-4-8(7)6-9/h1-4H,5-6H2

InChI key

UMJJFEIKYGFCAT-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Indanone undergoes TiCl4-Mg mediated coupling with CHBr3 to yield dibromomethyl carbinol. It reacts with 5,5-dimethyl-3-pyrazolidinone to yield 5,5-dimethyl-2-(1H-indenyl-2)-3-pyrazolidinone. 2-Indanone on photolysis by 266-nm one-photon excitation yields o-xylylene.

Application

2-Indanone was used as starting reagent in the synthesis of indene-fused porphyrins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectroscopic studies on photochemical formation of o-xylylene in solution.
Fujiwara M, et al.
The Journal of Physical Chemistry A, 101(27), 4912-4915 (1997)
Tu-Hsin Yan et al.
Organic letters, 15(22), 5802-5805 (2013-11-07)
TiCl4-Mg can mediate addition of CHBr3 to a variety of aldehydes and ketones to form dibromomethyl carbinols and also be used to effect CBr3 transfer to carbonyl groups to form tribromomethyl carbinols. The successful application of TiCl4-Mg-promoted coupling of CHBr3
Timothy D Lash et al.
The Journal of organic chemistry, 76(13), 5335-5345 (2011-05-24)
Indene-fused porphyrins have been synthesized starting from 2-indanone. Knorr-type reaction of oximes derived from benzyl or tert-butyl acetoacetate with 2-indanone and zinc dust in propionic acid gave good yields of indenopyrroles. Treatment with N-chlorosuccinimide then gave 8-chloro derivatives, and these
Laura E Korhonen et al.
Journal of medicinal chemistry, 48(11), 3808-3815 (2005-05-27)
The purpose of this study was to determine the cytochrome P450 1A2 (CYP1A2) inhibition potencies of structurally diverse compounds to create a comprehensive three-dimensional quantitative structure-activity relationship (3D-QSAR) model of CYP1A2 inhibitors and to use this model to predict the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service