Skip to Content
Merck
All Photos(1)

Key Documents

137456

Sigma-Aldrich

2-Ethyl-2-oxazoline

≥99%

Synonym(s):

2-Ethyl-4,5-dihydrooxazole, 2-Ethyloxazoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO
CAS Number:
Molecular Weight:
99.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

form

liquid

refractive index

n20/D 1.437 (lit.)

bp

128.4 °C (lit.)

mp

−62 °C (lit.)

density

0.982 g/mL at 25 °C (lit.)

SMILES string

CCC1=NCCO1

InChI

1S/C5H9NO/c1-2-5-6-3-4-7-5/h2-4H2,1H3

InChI key

NYEZZYQZRQDLEH-UHFFFAOYSA-N

General description

2-Ethyl-2-oxazoline undergoes cationic ring opening graft polymerization using tosylated cellulose nanowhisker as macroinitiator.

Application

2-Ethyl-2-oxazoline (EtOx) was used in synthesis of block copolymers of EtOx and styrene by cationic ring-opening polymerization and atom transfer radical polymerization.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luqiao Wen et al.
Drug development and industrial pharmacy, 46(2), 283-295 (2020-01-17)
In this study, we evaluated and screened the effects of the molecular weight (MW) and molar ratio of poly(2-ethyl-2-oxazoline)-cholesteryl methyl carbonate (PEtOz-CHMC) on the pH sensitivity, stability, and antitumor efficacy of liposomes. The pH sensitivity of PEtOz-CHMC with different MWs
Steffen Czich et al.
Molecules (Basel, Switzerland), 25(21) (2020-11-05)
The main task of tissue engineering (TE) is to reproduce, replicate, and mimic all kinds of tissues in the human body. Nowadays, it has been proven useful in TE to mimic the natural extracellular matrix (ECM) by an artificial ECM
Věra Mazánková et al.
Polymers, 12(11) (2020-11-19)
Polyoxazoline thin coatings were deposited on glass substrates using atmospheric pressure plasma polymerization from 2-ethyl-2-oxazoline vapours. The plasma polymerization was performed in dielectric barrier discharge burning in nitrogen at atmospheric pressure. The thin films stable in aqueous environments were obtained
Blaise L Tardy et al.
Biomacromolecules, 20(3), 1421-1428 (2019-02-23)
Metal-phenolic network (MPN) coatings have generated increasing interest owing to their biologically inspired nature, facile fabrication, and near-universal adherence, especially for biomedical applications. However, a key issue in biomedicine is protein fouling, and the adsorption of proteins on tannic acid-based
Synthesis of poly (2-ethyl-2-oxazoline)-b-poly (styrene) copolymers via a dual initiator route combining cationic ring-opening polymerization and atom transfer radical polymerization.
Becer CR, et al.
Macromolecules, 41(14), 5210-5215 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service