Skip to Content
Merck
All Photos(2)

Key Documents

13373

Sigma-Aldrich

Benzyldodecyldimethylammonium bromide

≥99.0% (AT)

Synonym(s):

Benzyldimethyldodecylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11N(Br)(CH3)2CH2C6H5
CAS Number:
Molecular Weight:
384.44
Beilstein:
3579193
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

description

cationic

Assay

≥99.0% (AT)

form

solid

mol wt

384.44 g/mol

SMILES string

[Br-].CCCCCCCCCCCC[N+](C)(C)Cc1ccccc1

InChI

1S/C21H38N.BrH/c1-4-5-6-7-8-9-10-11-12-16-19-22(2,3)20-21-17-14-13-15-18-21;/h13-15,17-18H,4-12,16,19-20H2,1-3H3;1H/q+1;/p-1

InChI key

KHSLHYAUZSPBIU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mikrocalorimetric studies on the interaction of N-dodecyl-N,N-dimethyl-N-benzylammonium halides with phosphatidylcholine bilayers: a counterion effect.
Bozenna Rózycka-Roszak et al.
Cellular & molecular biology letters, 7(2), 298-298 (2002-07-05)
Sterol and sterolin-containing products: hematologic adverse reactions.
Scott Jordan et al.
CMAJ : Canadian Medical Association journal = journal de l'Association medicale canadienne, 170(8), 1347-1348 (2004-04-28)
Bozenna Rózycka-Roszak et al.
Chemistry and physics of lipids, 123(2), 209-221 (2003-04-15)
The interaction of N-dodecyl-N,N-dimethyl-N-benzylammonium halides (DBeAX) with two types of phospholipid vesicles (MLV and SUV) was investigated using DSC and 1H NMR. It was suggested that the benzyl group like the micellisation process (J. Colloid Interface Sci. 218 (1999) 529)
M Rózalska et al.
Medycyna doswiadczalna i mikrobiologia, 45(3), 373-378 (1993-01-01)
The studies were performed at the Intensive Care Unit of the Institute of Pediatrics. There were aimed at determination of cleanliness of boxes together with ante-rooms. At the same time bacterial flora of hospitalized children was investigated. The bacteria were
H Benalla et al.
Journal of colloid and interface science, 272(2), 253-261 (2004-03-19)
Titration calorimetry has been used to study the effect of the addition of two primary alcohols, 1-butanol and 1-heptanol, to the aqueous phase on the thermal effects of micellization of benzyldimethyldodecylammonium bromide (BDDAB) as well as its adsorption onto nonporous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service