Skip to Content
Merck
All Photos(1)

Key Documents

133221

Sigma-Aldrich

Bis(5-chloro-2-hydroxyphenyl)methane

95%

Synonym(s):

Dichlorophen, Dichlorophene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2[C6H3(Cl)OH]2
CAS Number:
Molecular Weight:
269.12
Beilstein:
1884514
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

powder

mp

168-172 °C (lit.)

solubility

95% ethanol: soluble 1g/g
methanol: soluble
petroleum ether: soluble
toluene: very slightly soluble
water: insoluble

functional group

chloro

SMILES string

Oc1ccc(Cl)cc1Cc2cc(Cl)ccc2O

InChI

1S/C13H10Cl2O2/c14-10-1-3-12(16)8(6-10)5-9-7-11(15)2-4-13(9)17/h1-4,6-7,16-17H,5H2

InChI key

MDNWOSOZYLHTCG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(5-chloro-2-hydroxyphenyl)methane is a fungicide and is very effective against many celluloytic fungi, such as Penicillium, Aspergillus,Cladosporium and Trichoderma sp. It is a phenolic pesticide and undergoes photodegradation in the presence of visible-light absorber photosensitizer riboflavin.

Application

Bis(5-chloro-2-hydroxyphenyl)methane can be used as a starting material to synthesize methyl aluminium-(2,2′-methylene-p-chloro-bisphenoxide), which is used as a catalyst for the ring opening polymerization (ROP) of cyclic esters. It can also be used as an organic building block to prepare phosphomides substituted with aminoacid esters for various biological studies.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F J Verhagen et al.
Applied and environmental microbiology, 64(9), 3225-3231 (1998-09-03)
Higher fungi have a widespread capacity for biosynthesis of organohalogens. Commonly occurring chloroaromatic fungal metabolites can end up in anaerobic microniches at the boundary of fungal colonies and wetland soils. The aim of this study was to investigate the environmental
Reaction of methyl aluminium-(2, 2?-methylene-p-chloro-bisphenoxide) with 2, 2?-di (hydroxymethyl) biphenyl: A new aluminium complex bearing two kinds of diolate ligands
Ziemkowska W, et al.
Journal of Organometallic Chemistry, 693(3), 369-373 (2008)
Synthesis and antimicrobial activity of 2,10-dichloro-6-substituted amino acid ester12H-dibenzo[d,g][1,3,2]dioxaphosphocin-6-oxides
BS Kumar, et al.
Bulgarian Chemical Communications, 41(4), 422-426 (2009)
Antoine Ghauch et al.
Journal of hazardous materials, 164(2-3), 665-674 (2008-09-30)
Palladium, ruthenium and silver were investigated as catalysts for the dechlorination of dichlorophen (DCP, 2,2'-methylenebis(4-chlorophenol)), an antimicrobial and anthelmintic agent largely used as algicide, fungicide and bactericide. Experiments were undertaken under oxic and anoxic conditions for experimental durations up to
Juan P Escalada et al.
Journal of hazardous materials, 186(1), 466-472 (2010-12-07)
Within the context of environmentally friendly methods for the elimination of surface-water pollutants, the photodegradation of the phenolic pesticides bromoxynil (BXN) and dichlorophen (DCP) under simulated natural conditions has been studied. The work was done in the presence of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service