Skip to Content
Merck
All Photos(2)

Key Documents

133159

Sigma-Aldrich

Bathophenanthroline

greener alternative

97%

Synonym(s):

4,7-Diphenyl-1,10-phenanthroline, BPhen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16N2
CAS Number:
Molecular Weight:
332.40
Beilstein:
261048
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

218-220 °C (lit.)

Orbital energy

HOMO 6.4 eV 
LUMO 3 eV 

OLED Device Performance

ITO/MoO3/NPD/Alq3/BPhen/LiF/Al

  • Color: green
  • Max. Luminance: 20000 Cd/m2
  • Max. EQE: 1.2 %
  • Turn-On Voltage: 2.8 V

ITO/NPD/Alq3/BPhen/LiF/Al
  • Color: green
  • Max. Luminance: 19000 Cd/m2
  • Max. EQE: 1.16 %
  • Turn-On Voltage: 6.6 V

ITO/PEDOT:PSS/NPD/Alq3/BPhen/LiF/Al
  • Color: green
  • Max. Luminance: 14000 Cd/m2
  • Max. EQE: 1.44 %
  • Turn-On Voltage: 5 V

ITO/PEDOT:PSS/PVK:Ir(mppy)2 (6wt%)/BPhen/LiF:Al
  • Color: green
  • Max. Luminance: 10000 Cd/m2
  • Max. EQE: 1.1 %
  • Turn-On Voltage: 3.8 V

ITO/TCTA/Ir(ppy)3/BPhen/LiF/Al
  • Color: green
  • Max. Luminance: 300000 Cd/m2
  • Max. EQE: 19 %
  • Turn-On Voltage: 2.6 V

PEDOT:PSS/MoO3/NPD/Alq3/BPhen/LiF/Al
  • Color: green
  • Max. Luminance: 15000 Cd/m2
  • Max. EQE: 1.71 %
  • Turn-On Voltage: 2.3 V

greener alternative category

SMILES string

c1ccc(cc1)-c2ccnc3c2ccc4c(ccnc34)-c5ccccc5

InChI

1S/C24H16N2/c1-3-7-17(8-4-1)19-13-15-25-23-21(19)11-12-22-20(14-16-26-24(22)23)18-9-5-2-6-10-18/h1-16H

InChI key

DHDHJYNTEFLIHY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalysis. For more information see the Green, Catalytic Oxidation of Alcohols in Water

Application

  • Synthesis and luminescent properties of a ternary erbium (III) complex: A study on the luminescent properties of an erbium(III) complex with bathophenanthroline, which may have implications for materials science and sensor applications (Martín-Ramos et al., 2015).
  • Aggregation induced emission enhancement from Bathophenanthroline microstructures: Investigation of bathophenanthroline microstructures that exhibit enhanced emission properties and potential use as sensors for detecting mercury ions in water (Mazumdar et al., 2014).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The Journal of Organic Chemistry, 58, 7421-7421 (1993)
Nur Fadilah Jamaludin et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 19(9), 1075-1080 (2018-01-04)
Metal halide perovskites have demonstrated breakthrough performances as absorber and emitter materials for photovoltaic and display applications respectively. However, despite the low manufacturing cost associated with solution-based processing, the propensity for defect formation with this technique has led to an
Analytica Chimica Acta, 268, 351-351 (1992)
Katja Witzel et al.
Journal of proteome research, 6(4), 1325-1333 (2007-04-10)
This paper describes the application of the recently introduced fluorescence stain Ruthenium(II)-tris-(bathophenanthroline-disulphonate) (RuBP) on a comparative proteome analysis of two phenotypically different barley lines. We carried out an analysis of protein patterns from 2-D gels of the parental lines of
Inorganic Chemistry, 32, 3836-3836 (1993)

Articles

Since their discovery, organic light emitting devices (OLEDs) have evolved from a scientific curiosity into a technology with applications in flat panel displays and the potential to revolutionize the lighting market. During their relatively short history, the technology has rapidly advanced, and device efficiencies have increased more than 20-fold, approaching the theoretical limit for internal quantum efficiencies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service