Skip to Content
Merck
All Photos(1)

Key Documents

12804

Sigma-Aldrich

HBTU

≥98.0% (T), for peptide synthesis

Synonym(s):

N,N,N′,N′-Tetramethyl-O-(1H-benzotriazol-1-yl)uronium hexafluorophosphate, O-(Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16F6N5OP
CAS Number:
Molecular Weight:
379.24
Beilstein:
7328329
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

HBTU, ≥98.0% (T)

Assay

≥98.0% (T)

form

solid

reaction suitability

reaction type: Coupling Reactions

mp

200 °C (dec.) (lit.)

solubility

acetonitrile: 0.1 g/mL, clear

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

SMILES string

F[P-](F)(F)(F)(F)F.CN(C)C(\On1nnc2ccccc12)=[N+](/C)C

InChI

1S/C11H16N5O.F6P/c1-14(2)11(15(3)4)17-16-10-8-6-5-7-9(10)12-13-16;1-7(2,3,4,5)6/h5-8H,1-4H3;/q+1;-1

InChI key

UQYZFNUUOSSNKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

HBTU is a peptide coupling agent used in Fmoc-based solid-phase peptide synthesis.[1]

Application

HBTU is a peptide coupling agent for peptide synthesis.[2] It has also been used in standard Fmoc-based solid-phase peptide synthesis (SPPS) protocol.[3]
Recent studies show that crystal as well as solution structure of this coupling agent is a guanidinium N-oxide and not an uronium compound; Coupling reagent for peptide synthesis; advantages are: very low racemization, simple reaction conditions, very short reaction time, and high yields.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaido Kurrikoff et al.
Scientific reports, 7(1), 17056-17056 (2017-12-08)
Non-viral gene delivery systems have gained considerable attention as a promising alternative to viral delivery to treat diseases associated with aberrant gene expression. However, regardless of extensive research, only a little is known about the parameters that underline in vivo
Allan Bastos Lima et al.
Bioorganic & medicinal chemistry, 23(17), 5748-5755 (2015-08-04)
Dengue virus (DENV) and West Nile virus (WNV) are mosquito-borne arboviruses responsible for causing acute systemic diseases and severe health conditions in humans. The discovery of therapies capable to prevent infections or treat infected individuals remains an important challenge, since
Structural studies of reagents for peptide bond formation: Crystal and molecular structures of HBTU and HATU.
Abdelmoty I, et al.
Lett. Pept. Sci., 1(2), 57-67 (1994)
The uronium/guanidinium peptide coupling reagents: finally the true uronium salts.
Carpino LA, et al.
Angewandte Chemie (International Edition in English), 41(3), 441-445 (2002)
Hiroshi Wada et al.
PloS one, 13(7), e0199249-e0199249 (2018-07-04)
Recently, many autologous tumor antigens have been examined for their potential use in cancer immunotherapy. However, the success of cancer vaccines in clinical trials has been limited, partly because of the limitations of using single, short peptides in most attempts.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service