Skip to Content
Merck
All Photos(1)

Key Documents

126209

Sigma-Aldrich

2,3-Dihydroxybenzoic acid

99%

Synonym(s):

Hypogallic acid, Pyrocatechuic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
Beilstein:
2209117
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

204-206 °C (lit.)

solubility

methanol: soluble 50 mg/mL, clear to faintly hazy, orange

functional group

carboxylic acid

SMILES string

OC(=O)c1cccc(O)c1O

InChI

1S/C7H6O4/c8-5-3-1-2-4(6(5)9)7(10)11/h1-3,8-9H,(H,10,11)

InChI key

GLDQAMYCGOIJDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,3-Dihydroxybenzoic acid was used to study the complexes of manganese with aliphatic and aromatic polyhydroxy ligands in basic media by electrochemical, spectrophotometric and magnetic methods. It was used in isolation of new siderophore named vulnibactin from low iron cultures of Vibrio vulnificus, a human pathogen. It was used as cocrystal former to study the influence of position isomerism on the co-crystals formation and physicochemical properties of piracetam.

Biochem/physiol Actions

2,3-Dihydroxybenzoic acid was found to be orally effective iron-chelating drug in hypertransfused rat model. It is monocatechol siderophore produced by Brucella abortus.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nao Yamakawa et al.
Nature communications, 9(1), 4647-4647 (2018-11-09)
The emergence of zebrafish Danio rerio as a versatile model organism provides the unique opportunity to monitor the functions of glycosylation throughout vertebrate embryogenesis, providing insights into human diseases caused by glycosylation defects. Using a combination of chemical modifications, enzymatic
François Le Mauff et al.
The Journal of biological chemistry, 294(28), 10760-10772 (2019-06-07)
During infection, the fungal pathogen Aspergillus fumigatus forms biofilms that enhance its resistance to antimicrobials and host defenses. An integral component of the biofilm matrix is galactosaminogalactan (GAG), a cationic polymer of α-1,4-linked galactose and partially deacetylated N-acetylgalactosamine (GalNAc). Recent
The identification of 2, 3-dihydroxybenzoic acid as a potentially useful iron-chelating drug.
J H Graziano et al.
The Journal of pharmacology and experimental therapeutics, 190(3), 570-575 (1974-09-01)
Anna Nilsson et al.
Scientific reports, 7(1), 6352-6352 (2017-07-27)
Knowledge about the region-specific absorption profiles from the gastrointestinal tract of orally administered drugs is a critical factor guiding dosage form selection in drug development. We have used a novel approach to study three well-characterized permeability and absorption marker drugs
N Okujo et al.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine, 7(2), 109-116 (1994-04-01)
A new siderophore named vulnibactin has been isolated from low iron cultures of Vibrio vulnificus, a human pathogen. The structure was established as N-[3-(2,3-dihydroxybenzamido)propyl]-1,3-bis[2-(2-hydroxy-phenyl)- trans-5 - methyl-2-oxazoline-4-carboxamido]propane by a combination of acid hydrolysis, nuclear magnetic resonance spectroscopy and positive fast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service