Skip to Content
Merck
All Photos(1)

Key Documents

125032

Sigma-Aldrich

Chlorodiphenylmethane

98%

Synonym(s):

Benzhydryl chloride, Diphenylchloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2CHCl
CAS Number:
Molecular Weight:
202.68
Beilstein:
607925
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.595 (lit.)

bp

140 °C/3 mmHg (lit.)

mp

15-17 °C (lit.)

density

1.14 g/mL at 25 °C (lit.)

functional group

chloro
phenyl

SMILES string

ClC(c1ccccc1)c2ccccc2

InChI

1S/C13H11Cl/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,13H

InChI key

ZDVDCDLBOLSVGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chlorodiphenylmethane was used as initiator during the controlled radical polymerization of styrene catalyzed by ionic iron complex. It was also used as the starting reagent in the synthesis of trimethylhydroquinone derivatives.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Kawasaki et al.
Chemical & pharmaceutical bulletin, 47(2), 177-181 (1999-03-11)
A novel series of trimethylhydroquinone derivatives was synthesized and evaluated for their anti-lipid peroxidation activity in rat liver microsomes, inhibition of rat basophilic leukemia-1 (RBL-1) cell 5-lipoxygenase and 48 h homologous passive cutaneous anaphylaxis (PCA) activity in rats. 4-[4-[4-(Diphenylmethyl)-1-piperazinyl]-butoxy]-2,3,6-trimethyl phenol
Shota Niibayashi et al.
Chemical communications (Cambridge, England), (18)(18), 1855-1857 (2007-05-04)
Ionic iron complex [(Me(3)tacn)(2)Fe(2)Cl(3)](+)[(Me(3)tacn)FeCl(3)](-) (1), which is readily soluble in methanol, acted as a powerful catalyst in controlled radical polymerization of styrene and MMA, and showed promising features of removal from the resulting polymers and was reusable after recovery from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service