Skip to Content
Merck
All Photos(1)

Key Documents

118354

Sigma-Aldrich

9-Decen-1-ol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CH(CH2)8OH
CAS Number:
Molecular Weight:
156.27
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39020322
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.447 (lit.)

bp

234-238 °C (lit.)

density

0.876 g/mL at 25 °C (lit.)

SMILES string

OCCCCCCCCC=C

InChI

1S/C10H20O/c1-2-3-4-5-6-7-8-9-10-11/h2,11H,1,3-10H2

InChI key

QGFSQVPRCWJZQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

9-Decen-1-ol undergoes oxidation to give 9-decenoic in aqueous media (dioxane–water mixtures) in the presence of supported palladium or platinum catalysts.

Application

9-Decen-1-ol was used in the preparation of semifluorinated acids required for the synthesis of poly(styrene-b-semifluorinated isoprene) block copolymers with -CF2H-terminated side groups.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Oxidation of 9-decen-1-ol (rosalva) by air in aqueous media on platinum catalysts.
Crozon AB, et al.
New. J. Chem., 22(3), 269-273 (1998)
Effect of changing molecular end groups on surface properties: synthesis and characterization of poly (styrene-b-semifluorinated isoprene) block copolymers with-CF2H end groups.
Hayakawa T, et al.
Macromolecules, 33(21), 8012-8019 (2000)
Erik Selander et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(20), 6395-6400 (2015-04-29)
Interactions among microscopic planktonic organisms underpin the functioning of open ocean ecosystems. With few exceptions, these organisms lack advanced eyes and thus rely largely on chemical sensing to perceive their surroundings. However, few of the signaling molecules involved in interactions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service