Skip to Content
Merck
All Photos(3)

Documents

117323

Sigma-Aldrich

4,4′-Methylene-bis(2-chloroaniline)

85%

Synonym(s):

2,2′-Dichloro-4,4′-methylenedianiline, 4,4′-Diamino-3,3′-dichlorodiphenylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2[C6H3(Cl)NH2]2
CAS Number:
Molecular Weight:
267.15
Beilstein:
1882318
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

85%

form

solid

bp

202-214 °C/0.3 mmHg (lit.)

mp

102-107 °C (lit.)

SMILES string

Nc1ccc(Cc2ccc(N)c(Cl)c2)cc1Cl

InChI

1S/C13H12Cl2N2/c14-10-6-8(1-3-12(10)16)5-9-2-4-13(17)11(15)7-9/h1-4,6-7H,5,16-17H2

InChI key

IBOFVQJTBBUKMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Luukkanen et al.
Pharmacology & toxicology, 80(3), 152-158 (1997-03-01)
Rats were treated with acetone, pyrazole, phenobarbital, 4,4'-methylenebis-(2-chloroaniline) (MOCA), 3-methylcholanthrene, creosote oil, or a mixture of polychlorinated biphenyls (Aroclor 1254) to study the inducibility and enzyme kinetics of UDP-glucuronosyltransferases towards 1-hydroxypyrene, which is a human metabolite and a urinary biomarker
M Schuler et al.
Mutation research, 392(1-2), 81-95 (1997-08-01)
The in vitro micronucleus assay in conjunction with CREST-staining and fluorescence in situ hybridization (FISH) with centromere-specific DNA probes is being increasingly utilized for the detection of clastogenic and aneuploidy-inducing agents. Although potentially powerful techniques, both methods have unique characteristics
Hong-I Chen et al.
Journal of occupational health, 49(5), 389-398 (2007-10-24)
Oxidative DNA damage may play an important role in the human carcinogenic process. Recently, we reported a case of bladder cancer among 4, 4'-methylenebis (2-chloroaniline) (MBOCA)-exposed workers. By measuring the plasma level of 8-hydroxydeoxyguanosine (8-OHdG), we investigated the association between
4,4'-Methylenebis(2-chloroaniline).
Report on carcinogens : carcinogen profiles, 11, III167-III168 (2004-01-01)
E B Murray et al.
Reviews on environmental health, 20(3), 163-176 (2005-12-13)
Cytogenetic end-points used to estimate risk of genotoxic events in workers include the measurement of micronuclei (MN) in exfoliated cells, lymphocytes, and other tissues. Micronuclei are chromatin-containing bodies outside the cell nucleus resulting from contaminant-induced DNA damage. A review of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service