Skip to Content
Merck
All Photos(2)

Documents

112089

Sigma-Aldrich

(−)-β-Pinene

99%

Synonym(s):

(-)-beta-Pinene, (1S)-(−)-β-Pinene, (1S,5S)-2(10)-Pinene, (1S,5S)-6,6-Dimethyl-2-methylenebicyclo[3.1.1]heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
2038282
EC Number:
MDL number:
UNSPSC Code:
12352212
eCl@ss:
39011007
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.7 (vs air)

vapor pressure

~2 mmHg ( 20 °C)

Assay

99%

optical activity

[α]20/D −21°, neat

refractive index

n20/D 1.478 (lit.)

bp

165-167 °C (lit.)

mp

−61 °C (lit.)

density

0.859 g/mL at 25 °C

SMILES string

[H][C@]12CCC(=C)[C@]([H])(C1)C2(C)C

InChI

1S/C10H16/c1-7-4-5-8-6-9(7)10(8,2)3/h8-9H,1,4-6H2,2-3H3/t8-,9-/m0/s1

InChI key

WTARULDDTDQWMU-IUCAKERBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(-)-β-Pinene belongs to the terpenes family and is used as a starting material or intermediate in organic synthesis.

Application

(-)-β-Pinene has been used as test compound to assess its fumigant toxicity against all life stages of confused flour beetle and Mediterranean flour moth, via single-dose test. It has been used as a standard during the determination of constituents of the essential oil of Achillea ligustica by gas chromatography-mass spectrometry. (-)-β-Pinene can be used as a starting material to synthesize (1R,2S,3R,5R)-3-amino-6,6-dimethyl-2-hydroxybicyclo[3.1.1]heptane and (+)-nopinone.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral terpene auxiliaries. Part 1: Highly enantioselective reduction of ketones with borane catalyzed by an oxazaborolidine derived from (-)-?-pinene
Krzeminski MP and Wojtczak A
Tetrahedron Letters, 46(48), 8299-8302 (2005)
Fumigant toxicity of monoterpenoid compounds against the confused flour beetle, Tribolium confusum Jacquelin du Val.(Coleoptera: Tenebrionidae).
Saglam O and Ozder N.
Turkiye Entomoloji Dergisi (Turkish Journal of Entomology), 37(4) (2013)
Synthesis of (+)-Methyl Phaseate and Its Isomer from (-)-?-Pinene
Takahashi S, et al.
Agricultural and Biological Chemistry, 53(10), 2711-2718 (1989)
Dual stereoselectivity in the dialkylzinc reaction using (-)-?-pinene derived amino alcohol chiral auxiliaries
Binder CM, et al
The Journal of Organic Chemistry, 74(6), 2337-2343 (2009)
Platinum/tin catalyzed hydroformylation of naturally occurring monoterpenes
Gusevskaya EV, et al
J. Mol. Catal. A: Chem., 152(1), 15-24 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service