Skip to Content
Merck
All Photos(3)

Key Documents

108383

Sigma-Aldrich

1,1,3,3-Tetramethoxypropane

99%

Synonym(s):

Malonaldehyde bis(dimethyl acetal)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2CHCH2CH(OCH3)2
CAS Number:
Molecular Weight:
164.20
Beilstein:
1700020
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021012
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.407 (lit.)

bp

183 °C (lit.)
70-75 °C/15 mmHg (lit.)

density

0.997 g/mL at 25 °C (lit.)

functional group

acetal
ether

SMILES string

COC(CC(OC)OC)OC

InChI

1S/C7H16O4/c1-8-6(9-2)5-7(10-3)11-4/h6-7H,5H2,1-4H3

InChI key

XHTYQFMRBQUCPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

1,1,3,3-Tetramethoxypropane undergoes acid hydrolysis to yield malondialdehyde.

Application

1,1,3,3-Tetramethoxypropane was used in preparing malondialdehyde standards during malondialdehyde assay on lung homogenates. It was used to study the effect of Salvianolic acid A on the migration and proliferation of vascular smooth muscle cells.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

134.6 °F - closed cup - DIN 51755 Part 1

Flash Point(C)

57 °C - closed cup - DIN 51755 Part 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masoud Fallah Rajabpour Zare et al.
Life sciences, 232, 116623-116623 (2019-07-08)
Doxorubicin, an antibiotic belonging to anthracycline family, has been used for treatment of malignancies. Cardiotoxicity is the main adverse effect of doxorubicin. Apigenin, as a flavonoid, has antioxidant, anti-inflammatory and anti-tumoral properties. The aim of this study was the assessment
Sihem Dabbou et al.
Meat science, 124, 77-83 (2016-11-14)
A trial was performed to evaluate bilberry pomace (BP) as alternative source of nutrients for rabbits. One hundred and forty-four Grimaud weaned rabbits were divided into 4 groups of 36 animals each and fed ad libitum with a basal diet
Laura Gasco et al.
Animals : an open access journal from MDPI, 9(9) (2019-09-01)
This trial investigated the effect of the dietary inclusion of Hermetia illucens (H) and Tenebrio molitor (T) fats as alternative lipid sources for growing rabbits, and assessed the carcass characteristics; proximate composition; lipid peroxidation, and fatty acid profile of the
Bleomycin-induced strand-scission of DNA. Mechanism of deoxyribose cleavage.
L Giloni et al.
The Journal of biological chemistry, 256(16), 8608-8615 (1981-08-25)
Foula Sozo et al.
American journal of physiology. Lung cellular and molecular physiology, 296(3), L510-L518 (2008-12-30)
Little is known about the effects of fetal ethanol exposure on lung development. Our aim was to determine the effects of repeated ethanol exposure during late gestation on fetal lung growth, maturation, and inflammatory status. Pregnant ewes were chronically catheterized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service