Skip to Content
Merck
All Photos(3)

Key Documents

108030

Sigma-Aldrich

3-Chloro-2-methyl-1-propene

technical grade, 90%

Synonym(s):

β-Methallyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)CH2Cl
CAS Number:
Molecular Weight:
90.55
Beilstein:
878160
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050134
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

vapor density

3.12 (vs air)

vapor pressure

102 mmHg ( 20 °C)

Assay

90%

autoignition temp.

899 °F

contains

~600 ppm BHT as stabilizer

expl. lim.

9.3 %

refractive index

n20/D 1.427 (lit.)

bp

71-72 °C (lit.)

mp

−80 °C (lit.)

solubility

H2O: insoluble
acetone: soluble
chloroform: soluble
diethyl ether: soluble
ethanol: soluble

density

0.925 g/mL at 20 °C (lit.)

functional group

alkyl halide
chloro

storage temp.

2-8°C

SMILES string

CC(=C)CCl

InChI

1S/C4H7Cl/c1-4(2)3-5/h1,3H2,2H3

InChI key

OHXAOPZTJOUYKM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Chloro-2-methyl-1-propene is the reactant for the synthesis of cyclobutanone.

Application

3-Chloro-2-methyl-1-propene was used to study ring opening polymerization of oxirane derivatives using organotin phosphate condensate. It was used to study cationic polymerization of 3-Chloro-2-methyl-1-propene using using AICI3 and A3IBr, as initiators.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The cationic polymerization of 3-chloro-2-methyl-1-propene.
Marek M, et al.
Makromol. Chem., 187(10), 2337-2344 (1986)
Cyclobutanone from Methylenecyclopropane via Oxaspiropentane.
Salaun JRJC and Conia JM.
Organic Syntheses, 36-36 (1977)
3-Chloro-2-methylpropene.
Report on carcinogens : carcinogen profiles, 11, III57-III58 (2004-01-01)
Ring-opening polymerization of various oxirane derivatives using organotin phosphate condensate; selective synthesis of the polyether containing oxirane ring in the side chain.
Iwasa N, et al.
Polym. Bull., 61(2), 207-216 (2008)
K Hooper et al.
American journal of industrial medicine, 22(6), 793-808 (1992-01-01)
In California, 370 carcinogens and 112 reproductive/developmental toxicants have been identified as a result of the State's Safe Drinking Water and Toxic Enforcement Act of 1986. They include pesticides, solvents, metals, industrial intermediates, environmental mixtures, and reactive agents. Occupational, environmental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service