Skip to Content
Merck
All Photos(1)

Documents

106879

Sigma-Aldrich

1,1′-Diacetylferrocene

97%

Synonym(s):

Bis(acetylcyclopentadienyl)iron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14FeO2
CAS Number:
Molecular Weight:
270.10
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:

Assay

97%

reaction suitability

core: iron
reagent type: catalyst

mp

122-128 °C

SMILES string

[Fe].CC(=O)[C]1[CH][CH][CH][CH]1.CC(=O)[C]2[CH][CH][CH][CH]2

InChI

1S/2C7H7O.Fe/c2*1-6(8)7-4-2-3-5-7;/h2*2-5H,1H3;

InChI key

JNBIZDIDVCJECQ-UHFFFAOYSA-N

General description

1,1′-Diacetylferrocene is an organic compound used as a catalyst or ligand in certain cross-coupling reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yulia P Tupolova et al.
Dalton transactions (Cambridge, England : 2003), 49(44), 15592-15596 (2020-11-04)
Herein, we report the synthesis and magnetic properties of the Co(ii) coordination compound with the 1,1'-bis(1-((pyrid-2-ylmethylene)hydrazono)ethyl)ferrocene (L) ligand, having the general formula [CoLCl2]. The static magnetic data analysis supported by the CASSCF/NEVPT2 calculations revealed the presence of the triaxial magnetic
Formation of functionalized [3] ferrocenophane derivatives by an enamine condensation reaction
Knuppel S, et al.
Journal of Organometallic Chemistry, 586, 218-222 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service