Merck
  • A covalently bound inhibitor triggers EZH2 degradation through CHIP-mediated ubiquitination.
A covalently bound inhibitor triggers EZH2 degradation through CHIP-mediated ubiquitination.
The EMBO journal (2017-03-23)
Xu Wang, Wei Cao, Jianjun Zhang, Ming Yan, Qin Xu, Xiangbing Wu, Lixin Wan, Zhiyuan Zhang, Chenping Zhang, Xing Qin, Meng Xiao, Dongxia Ye, Yuyang Liu, Zeguang Han, Shaomeng Wang, Li Mao, Wenyi Wei, Wantao Chen
ABSTRACT
Enhancer of zeste homolog 2 (EZH2) has been characterized as a critical oncogene and a promising drug target in human malignant tumors. The current EZH2 inhibitors strongly suppress the enhanced enzymatic function of mutant EZH2 in some lymphomas. However, the recent identification of a PRC2- and methyltransferase-independent role of EZH2 indicates that a complete suppression of all oncogenic functions of EZH2 is needed. Here, we report a unique EZH2-targeting strategy by identifying a gambogenic acid (GNA) derivative as a novel agent that specifically and covalently bound to Cys668 within the EZH2-SET domain, triggering EZH2 degradation through COOH terminus of Hsp70-interacting protein (CHIP)-mediated ubiquitination. This class of inhibitors significantly suppressed H3K27Me3 and effectively reactivated polycomb repressor complex 2 (PRC2)-silenced tumor suppressor genes. Moreover, the novel inhibitors significantly suppressed tumor growth in an EZH2-dependent manner, and tumors bearing a non-GNA-interacting C668S-EZH2 mutation exhibited resistance to the inhibitors. Together, our results identify the inhibition of the signaling pathway that governs GNA-mediated destruction of EZH2 as a promising anti-cancer strategy.
Product Description
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys9) Antibody, clone 6F12-H4, clone 6F12-H4, from mouse