Merck
  • G9a drives hypoxia-mediated gene repression for breast cancer cell survival and tumorigenesis.
G9a drives hypoxia-mediated gene repression for breast cancer cell survival and tumorigenesis.
Proceedings of the National Academy of Sciences of the United States of America (2017-06-21)
Francesco Casciello, Fares Al-Ejeh, Greg Kelly, Donal J Brennan, Shin Foong Ngiow, Arabella Young, Thomas Stoll, Karolina Windloch, Michelle M Hill, Mark J Smyth, Frank Gannon, Jason S Lee
ABSTRACT
G9a is an epigenetic regulator that methylates H3K9, generally causing repression of gene expression, and participates in diverse cellular functions. G9a is genetically deregulated in a variety of tumor types and can silence tumor suppressor genes and, therefore, is important for carcinogenesis. Although hypoxia is recognized to be an adverse factor in tumor growth and metastasis, the role of G9a in regulating gene expression in hypoxia has not been described extensively. Here, we show that G9a protein stability is increased in hypoxia via reduced proline hydroxylation and, hence, inefficient degradation by the proteasome. This inefficiency leads to an increase in H3K9me2 at its target promoters. Blocking the methyltransferase activity of G9a inhibited cellular proliferation and migration in vitro and tumor growth in vivo. Furthermore, an increased level of G9a is a crucial factor in mediating the hypoxic response by down-regulating the expression of specific genes, including ARNTL, CEACAM7, GATA2, HHEX, KLRG1, and OGN This down-regulation can be rescued by a small molecule inhibitor of G9a. Based on the hypothesis that the changes in gene expression would influence patient outcomes, we have developed a prognostic G9a-suppressed gene signature that can stratify breast cancer patients. Together, our findings provide an insight into the role G9a plays as an epigenetic mediator of hypoxic response, which can be used as a diagnostic marker, and proposes G9a as a therapeutic target for solid cancers.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
UNC0642, ≥98% (HPLC)
Sigma-Aldrich
Anti-Lamin A/C Antibody, clone 14, clone 14, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-SUV39H1 Histone Methyltransferase antibody produced in mouse, ~2 mg/mL, clone 44.1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Anti-G9a Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Cytoplasmic FMR1-interacting protein 1 Antibody, from rabbit, purified by affinity chromatography