Merck
  • Distinguishing hyperglycemic changes by Set7 in vascular endothelial cells.
Distinguishing hyperglycemic changes by Set7 in vascular endothelial cells.
Circulation research (2012-03-10)
Jun Okabe, Christian Orlowski, Aneta Balcerczyk, Chris Tikellis, Merlin C Thomas, Mark E Cooper, Assam El-Osta
ABSTRACT
Epigenetic changes are implicated in the persisting vascular effects of hyperglycemia. The precise mechanism whereby chromatin structure and subsequent gene expression are regulated by glucose in vascular endothelial cells remain to be fully defined. We have studied the molecular and functional mechanism whereby the Set7 methyltransferase associates with chromatin formation and histone methylation in vascular cells in response to current and previous exposure to glucose. To characterize the molecular and functional identity of the Set7 protein, we used vascular cells overexpressing or lacking Set7. Chromatin fractionation for mono-methylation of lysine 4 on histone H3 identified methyltransferase activity. Immunofluorescence experiments strongly suggest that Set7 protein accumulates in the nucleus in response to hyperglycemia. Moreover, activation of proinflammatory genes by high glucose is dependent on Set7 but distinguished by H3K4m1 gene patterns. We show that transient hyperglycemia regulates the expression of proinflammatory genes in vascular endothelial cells in vitro and the persistent increase in glucose-induced gene expression in the aorta of nondiabetic mice. This study uncovers that the response to hyperglycemia in vascular endothelial cells involves the H3K4 methyltransferase, Set7. This enzyme appears to regulate glucose-induced chromatin changes and gene expression not only by H3K4m1-dependent but also H3K4m1-independent pathways. Furthermore, Set7 appears to be responsible for sustained vascular gene expression in response to prior hyperglycemia and is a potential molecular mechanism for the phenomenon of hyperglycemic memory.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-hnRNP-A1 antibody, Mouse monoclonal, clone 9H10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-HP1α Antibody, clone15.19s2, clone 15.19s2, Upstate®, from mouse
Sigma-Aldrich
Anti-monomethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-c-Jun Antibody, serum, Upstate®
Sigma-Aldrich
Monoclonal Anti-SUV39H1 Histone Methyltransferase antibody produced in mouse, ~2 mg/mL, clone 44.1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-CREB Antibody, Upstate®, from rabbit