Skip to Content

06-863

Anti-CREB Antibody

Upstate®, from rabbit

Synonym(s):

Cyclic AMP-responsive element-binding protein 1, cAMP-responsive element-binding protein 1, CREB-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ChIP, EMSA, IP, WB
Citations:
145
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

using Protein A

species reactivity

mouse, human, rat

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), horse (based on 100% sequence homology), canine (based on 100% sequence homology)

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq), electrophoretic mobility shift assay: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CREB1(1385)

General description

Cyclic AMP-responsive element-binding protein 1 (UniProt P16220; also known as cAMP-responsive element-binding protein 1, CREB-1) is encoded by the CREB1 gene (Gene ID 1385) in human. CREB is a transcription factor that binds the cAMP response elements (CRE) and regulates the transcription of the downstream genes. CREB activity is regulation via phosphorylation by various kinases, including PKA, and Ca2+/calmodulin-dependent protein kinases. Genes known to be regulated by CREB include c-fos, BDNF, tyrosine hydroxylase, neuropeptides (such as somatostatin, enkephalin, VGF, and corticotropin-releasing hormone), as well as the circadian clock genes PER1 and PER2. CREB is critical for memory formation across a wide range of species.
~43 kDa observed.

Immunogen

Epitope: Amino acids 5-24.
KLH-conjugated, synthetic peptide (CSGAENQ QSGDAAVTEAENQQ) corresponding to a.a. 5-24 of human CREB. The immunizing sequence is identical in pig, shares 19 of 20 residues with mouse and bovine and 18 of 20 residues with rat.

Application

Anti-CREB Antibody, Cat. No. 06-863, is a highly specific rabbit Polyclonal antibody, that targets CREB and has been tested in Chromatin Immunoprecipitation (ChIP), ChIP-seq, Electrophoretic Mobility Shift Assay (EMSA), Immunoprecipitation, and Western Blotting.
Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated CREB in 500 µg of HeLa nuclear extract.
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected an enhanced CREB occupancy at the LYPD3 promoter in LKB1-deficient TE-4 human esophageal cancer cells (Gu, Y., et al. (2012). Oncogene. 31(4):469–479).
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected CREB association at the RNASET2 sites targeted by T-cell Leukemia Virus type 1 (HTLV-1) encoded Tax protein using HTLV-1-infected human T-cell lines, MT-2, C8166/45 and SLB-1 (Polakowski, N., et al. (2011). Viruses. 3(8):1485-500).
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected CREB occupancy at the hBDNF promoter IV using postmortem human parietal cortex tissue chromatin preparations (Pruunsild, P., et al. (2011). J. Neurosci. 31(9):3295-3308).
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected CREB occupancy at the miR-9-2 promoter in SK-N-BE human neuroblastoma cells (Laneve, P., et al. (2010). Nucl. Acids Res. 38(20):6895-6905).
ChIP-seq Analysis: A representative lot identified CREB target genes in murine spermatogonia derived GC1-spg cells by ChIP-seq analysis (Martianov, I., et al. (2010). BMC Genomics. 11:530).
Western Blotting Analysis: Representative lots detected CREB in lysates from cultured embryonic rat striatal neurons and PC12 pheochromocytoma cells (Hutchinson, A.N., et al. (2012). Neuropsychopharmacology. 37(2):321-337; Maharjan, S., et al. (2010). J. Neurochem. 112(1):42-55).
Western Blotting Analysis: Representative lots detected CREB in mouse proximal caput epididymis-1 (PC-1) cell lysate and hippocampus tissue homogenates (Hamzeh, M., and Robaire, B. (2011). J. Endocrinol. 209(1):55-64; Zhou, Z., et al. (2009). Neuropharmacology. 56(1):81-89).
Western Blotting Analysis: A representative lot detected CREB in SK-N-BE human neuroblastoma cell lysate (Laneve, P., et al. (2010). Nucl. Acids Res. 38(20):6895-6905).
Electrophoretic Mobility Shift Assay (EMSA): A representative lot caused a supershift of DNA-CREB complex in EMSA using radiolabeled oligonucleotide containing hBDNF promoter I CREB-binding sequence and lysates from KCl-treated primary rat neurons (Pruunsild, P., et al. (2011). J. Neurosci. 31(9):3295-3308).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Biochem/physiol Actions

This antibody recognizes CREB.

Physical form

Protein A purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide with 30% glycerol.

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.
Evaluated by Western Blotting in HeLa nuclear extract.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected CREB in 10 µg of HeLa nuclear extract.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 04-218

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Explore all of our products under Anti-CREB Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library