Merck
  • Biodegradable Film for the Targeted Delivery of siRNA-Loaded Nanoparticles to Vaginal Immune Cells.
Biodegradable Film for the Targeted Delivery of siRNA-Loaded Nanoparticles to Vaginal Immune Cells.
Molecular pharmaceutics (2015-06-24)
Jijin Gu, Sidi Yang, Emmanuel A Ho
ABSTRACT
The goal of this study was to develop and characterize a novel intravaginal film platform for targeted delivery of small interfering RNA (siRNA)-loaded nanoparticles (NP) to dendritic cells as a potential gene therapy for the prevention of sexually transmitted human immunodeficiency virus (HIV) infection. Poly(ethylene glycol) (PEG)-functionalized poly(D, L-lactic-co-glycolic acid) (PLGA)/polyethylenimine (PEI)/siRNA NP (siRNA-NP) were fabricated using a modified emulsion-solvent evaporation method and characterized for particle size, zeta potential, encapsulation efficiency (EE), and siRNA release. siRNA-NP were decorated with anti-HLA-DR antibody (siRNA-NP-Ab) for targeting delivery to HLA-DR+ dendritic cells (DCs) and homogeneously dispersed in a biodegradable film consisting of poly vinyl alcohol (PVA) and λ-carrageenan. The siRNA-NP-Ab-loaded film (siRNA-NP-Ab-film) was transparent, displayed suitable physicomechanical properties, and was noncytotoxic. Targeting activity was evaluated in a mucosal coculture model consisting of a vaginal epithelial monolayer (VK2/E6E7 cells) and differentiated KG-1 cells (HLA-DR+ DCs). siRNA-NP-Ab were rapidly released from the film and were able to penetrate the epithelial layer to be taken up by differentiated KG-1 cells. siRNA-NP-Ab demonstrated higher targeting activity and significantly higher knockdown of synaptosome-associated 23-kDa protein (SNAP-23) mRNA and protein when compared to siRNA-NP without antibody conjugation. Overall, these data suggest that our novel siRNA-NP-Ab-film may be a promising platform for preventing HIV infection within the female genital tract.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)
Sigma-Aldrich
EGF from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
Folate-poly(ethylene glycol)-b-poly(lactide-co-glycolide), PEG average Mn 2,000, PLGA average Mn 10,000, lactide:glycolide 50:50
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide)(85/15)-b-poly(ethylene glycol)-carboxylic acid, 5k-13k
Sigma-Aldrich
Carboxylic acid-poly(ethylene glycol)-b-poly(lactide-co-glycolide), PEG average Mn 5,000, PLGA average Mn 20,000, lactide:glycolide 50:50
Sigma-Aldrich
Biotin-poly(ethylene glycol)-b-poly(lactide-co-glycolide), PEG average Mn 2,000, PLGA average Mn 10,000, lactide:glycolide 50:50
Sigma-Aldrich
EGF from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
Sigma-Aldrich
Streptomycin Ready Made Solution, 100 mg/mL in water
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
EGF from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture