Merck
  • Cytosolic BolA Plays a Repressive Role in the Tolerance against Excess Iron and MV-Induced Oxidative Stress in Plants.
Cytosolic BolA Plays a Repressive Role in the Tolerance against Excess Iron and MV-Induced Oxidative Stress in Plants.
PloS one (2015-05-01)
Lu Qin, Meihuan Wang, Jia Zuo, Xiangyang Feng, Xuejiao Liang, Zhigeng Wu, Hong Ye
ABSTRACT
The BolA-like protein is present in all eukaryotes, and it is able to form complex with monothiol glutaredoxin of the same subcellular compartments, suggesting that the BolA-like protein has essential function in eukaryotes, and that the function is associated with its partner glutaredoxin. Some studies have indicated a role for BolA proteins in Fe-S cluster synthesis or in redox homeostasis. However, the physiological function of BolA proteins remains to be elucidated. Here, we report the characterization of an insertion mutant of BolA3 in Arabidopsis. Among the four AtBolA proteins found in Arabidopsis, the AtBolA3 was the only BolA located in the cytosol of plant cells. It was highly expressed in roots. AtBolA3 was able to interact with the cytosolic monothiol glutaredoxin, AtGRXS17. The bola3 mutant did not show any notable phenotype under normal growth condition, but rather grew better than wild type under some stresses. The bola3 mutant was more tolerant to excess iron and the MV-induced oxidative stress than wild type. It displayed no necrosis in leaves, developed longer roots, accumulated more iron and higher Fe-S protein activities in roots. In addition, the mutant possessed a more potent antioxidant defense to scavenge ROS species. Taken together, our data indicated that the cytosolic AtBolA3 has a suppressive role in the tolerance to excess iron and the MV-induced oxidative stress in plants. AtBolA3 seems to be a repressor under some stress conditions.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Scandium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Scandium, foil, 25x25mm, thickness 0.1mm, as rolled, 99%
Scandium, foil, not light tested, 25x25mm, thickness 0.0125mm, as rolled, 99%
Scandium, foil, 50x50mm, thickness 0.1mm, as rolled, 99%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)