Merck
Improved profiling of estrogen metabolites by orbitrap LC/MS.
Steroids (2014-12-30)
Xingnan Li, Adrian A Franke
ABSTRACT
Estrogen metabolites are important biomarkers to evaluate cancer risks and metabolic diseases. Due to their low physiological levels, a sensitive and accurate method is required, especially for the quantitation of unconjugated forms of endogenous steroids and their metabolites in humans. Here, we evaluated various derivatives of estrogens for improved analysis by orbitrap LC/MS in human serum samples. A new chemical derivatization reagent was applied modifying phenolic steroids to form 1-methylimidazole-2-sulfonyl adducts. The method significantly improves the sensitivity 2-100 fold by full scan MS and targeted selected ion monitoring MS over other derivatization methods including, dansyl, picolinoyl, and pyridine-3-sulfonyl products.
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
4-Hydroxyestrone, ≥90% (HPLC)
Sigma-Aldrich
4-Hydroxyestradiol
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
2-Methoxyestradiol, powder
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Methylimidazole-2-sulfonyl chloride, 95%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Pyridine-3-sulfonyl chloride, 95%
Sigma-Aldrich
Testosterone, ≥98%
Sigma-Aldrich
Dansyl chloride, suitable for fluorescence, BioReagent, ≥99.0% (HPLC)
Sigma-Aldrich
2-Methyl-6-nitrobenzoic anhydride, 97%
Sigma-Aldrich
2-Picolinic acid, ReagentPlus®, 99%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)