Merck
  • Screening protein isoforms predictive for cancer using immunoaffinity capture and fast LC-MS in PRM mode.
Screening protein isoforms predictive for cancer using immunoaffinity capture and fast LC-MS in PRM mode.
Proteomics. Clinical applications (2015-02-07)
Antoine Lesur, Lina Ancheva, Yeoun Jin Kim, Guy Berchem, Jan van Oostrum, Bruno Domon
ABSTRACT
We report an immunocapture strategy to extract proteins known to harbor driver mutations for a defined cancer type before the simultaneous assessment of their mutational status by MS. Such a method bypasses the sensitivity and selectivity issues encountered during the analysis of unfractionated complex biological samples. Fast LC separations using short nanobore columns hyphenated with a high-resolution quadrupole-orbitrap mass spectrometer have been devised to take advantage of fast MS cycle times in conjunction with sharp chromatographic peak widths to accelerate the sample analysis throughput. Such an analytical platform is well suited to analyze simple protein mixtures obtained after immunoaffinity enrichment. After establishing the technical performance of the platform, the method was applied to the quantitative profiling of cellular Ras and EGFR protein isoforms, as well as serum amyloid A isoforms in plasma. Immunoaffinity purification combined with fast LC-MS detection for the detection of driver mutations in tissue and tumor biomarkers in plasma samples can assist clinicians to select an optimal therapeutic intervention for patients.
Sigma-Aldrich
Anti-EGFR Antibody, clone 528 (Azide-free), clone 528, from mouse
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Anti-Pan-Ras Antibody, clone RAS 10, clone RAS 10, from mouse