Merck
  • Characteristics of Extended-Spectrum β-Lactamase-Producing Escherichia coli in Retail Meats and Shrimp at a Local Market in Vietnam.
Characteristics of Extended-Spectrum β-Lactamase-Producing Escherichia coli in Retail Meats and Shrimp at a Local Market in Vietnam.
Foodborne pathogens and disease (2015-06-26)
Quoc Phong Le, Shuhei Ueda, Thi Ngoc Hue Nguyen, Thi Van Khanh Dao, Thi Ai Van Hoang, Thi Thuy Nga Tran, Itaru Hirai, Tatsuya Nakayama, Ryuji Kawahara, Thai Hung Do, Quang Mai Vien, Yoshimasa Yamamoto
ABSTRACT
Contamination of food with multiantibiotic-resistant bacteria, particularly extended-spectrum β-lactamase (ESBL)-producing Enterobacteriaceae, is considered a potential source for the wide dissemination of ESBL-producing bacteria in communities. However, little is known about the extent of contamination of food with ESBL-producing bacteria in Vietnam. This study was conducted to assess the characteristics of ESBL-producing Escherichia coli isolated from retail meats and shrimp in Nha Trang, Vietnam. A total of 350 food samples (poultry [n=143], pork [n=147], and shrimp [n=60]) were purchased in July and November 2013 from a local market. ESBL-producing E. coli were isolated, and ESBL genotypes, phylogenetic groups, and antibiotic resistance profiles were determined. The prevalence of ESBL-producing E. coli in retail foods was 40.6%. β-Lactamase-encoding genes of the CTX-M-1 (50.7%), CTX-M-9 (41.5%), TEM (59.9%), and SHV (2.8%) groups were detected singly or in combination. The percentages of single ESBL isolates harboring CTX-M-1 or -9 plus TEM groups were 35.2% and 16.2%, respectively. B1 was the most prevalent phylogroup in ESBL isolates from pork (44.7%), poultry (55.9%), and shrimp (72.7%). B2 was the least prevalent (4.2% and 4.8% for pork and poultry isolates, respectively). The prevalence of multidrug resistance (MDR; resistance to ≥ 3 antimicrobial groups) in ESBL-producing E. coli isolated from food was 85.9%. This is the first report of the characteristics of ESBL-producing E. coli in retail foods in a local city in Vietnam. Our findings indicate that retail foods are contaminated with ESBL-producing E. coli, of which many were MDR. Further monitoring and public health efforts targeting food administration are needed to control the spread of ESBL-producing bacteria in communities.
Sigma-Aldrich
X-GlcA, reagent for selection of recombinant bacterial clones
Sigma-Aldrich
5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt, ≥98%
Sigma-Aldrich
5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt, tablet