Skip to Content

System maintenance is scheduled for 11 Sep, 08:00 PM - 12 Sep, 00:00 AM CST [12 Sep 03:00 AM to 07:00 AM CET] and will affect some web and offline transactions, including quotes, price checks, and order status inquiries. We apologize for any inconvenience.

Merck

Key Documents

Skip To

B6650

X-GlcA

reagent for selection of recombinant bacterial clones

Synonym(s):

5-Bromo-4-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt, X-GlcA, X-glucuronide CHA salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H13BrClNO7 · C6H13N
CAS Number:
Molecular Weight:
521.79
NACRES:
NA.85
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

sterility

non-sterile

assay

≥98% (TLC)

form

powder

technique(s)

nucleic acid detection: suitable

solubility

DMF: soluble

suitability

suitable for β-galactosidase test

storage temp.

−20°C

SMILES string

NC1CCCCC1.O[C@@H]2[C@@H](O)[C@@H](O[C@@H]([C@H]2O)C(O)=O)Oc3c[nH]c4ccc(Br)c(Cl)c34

InChI

1S/C14H13BrClNO7.C6H13N/c15-4-1-2-5-7(8(4)16)6(3-17-5)23-14-11(20)9(18)10(19)12(24-14)13(21)22;7-6-4-2-1-3-5-6/h1-3,9-12,14,17-20H,(H,21,22);6H,1-5,7H2/t9-,10-,11+,12-,14+;/m0./s1

InChI key

JXCKZXHCJOVIAV-CYRSAHDMSA-N

General description

X-GlcA is a chromogenic substrate for β-galactosidase, used to determine the presence or absence of a cloned DNA insert in bacteria growing on agar plates. X-GlcA is designed to replace X-Gal in blue-white selection of recombinant bacterial colonies with the lac+ phenotype.

Application

Chromogenic substrate for β-glucuronidase (GUS) gene detection.
Suitable for use in selection of recombinant bacterial colonies with the lac+ phenotype.
X-GlcA has been used in histochemical staining of root sections for light microscopic observation.

Biochem/physiol Actions

When X-GlcA is hydrolyzed by β-galactosidase, the resulting product will form a blue precipitate. Lac+ colonies grown in the presence of X-GlcA turn an intense blue color, allowing for easy differentiation between lac+ and lac- colonies.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B8049B9146S9811
description

reagent for selection of recombinant bacterial clones

description

tablet

description

≥98%

description

reagent for selection of recombinant bacterial clones

suitability

suitable for β-galactosidase test

suitability

-

suitability

suitable for identification of lac+ bacterial colonies

suitability

suitable for β-galactosidase test

technique(s)

nucleic acid detection: suitable

technique(s)

-

technique(s)

-

technique(s)

-

grade

Molecular Biology

grade

-

grade

-

grade

Molecular Biology

form

powder

form

tablet

form

powder

form

powder

solubility

DMF: soluble

solubility

-

solubility

DMF: methanol (1:1): soluble 50 mg/mL

solubility

DMF: 50 mg/mL

assay

≥98% (TLC)

assay

-

assay

≥98%

assay

≥95% (HPLC)




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters