Merck
  • Conversion of polar and non-polar algae oil lipids to fatty acid methyl esters with solid acid catalysts--A model compound study.
Conversion of polar and non-polar algae oil lipids to fatty acid methyl esters with solid acid catalysts--A model compound study.
Bioresource technology (2015-05-26)
Martta Asikainen, Tony Munter, Juha Linnekoski
ABSTRACT
Bio-based fuels are becoming more and more important due to the depleting fossil resources. The production of biodiesel from algae oil is challenging compared to terrestrial vegetable oils, as algae oil consists of polar fatty acids, such as phospholipids and glycolipids, as well as non-polar triglycerides and free fatty acids common in vegetable oils. It is shown that a single sulphonated solid acid catalyst can perform the esterification and transesterification reactions of both polar and non-polar lipids. In mild reaction conditions (60-70 °C) Nafion NR50 catalyst produces methyl palmitate (FAME) from the palmitic acid derivatives of di-, and tri-glyceride, free fatty acid, and phospholipid with over 80% yields, with the glycolipid derivative giving nearly 40% yields of FAME. These results demonstrate how the polar and non-polar lipid derivatives of algal oil can be utilised as feedstocks for biodiesel production with a single catalyst in one reaction step.
Sigma-Aldrich
Methyl palmitate, ≥97%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sorbitan monopalmitate
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Methyl palmitate, ≥99% (capillary GC)
Sigma-Aldrich
Dipalmitin, ≥99.0%
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Montmorillonite, K 10, powder
Sigma-Aldrich
Glyceryl tripalmitate, ≥85%
Sigma-Aldrich
Methyl pentadecanoate, ≥98.5%
Sigma-Aldrich
Glyceryl tripalmitate, ≥99%
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine, semisynthetic, ≥99%
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine, ≥99% (TLC)