Merck
  • Endoplasmic reticulum stress impairs insulin receptor signaling in the brains of obese rats.
Endoplasmic reticulum stress impairs insulin receptor signaling in the brains of obese rats.
PloS one (2015-05-16)
Lina Liang, Jing Chen, Libin Zhan, Xiaoguang Lu, Xiaoxin Sun, Hua Sui, Luping Zheng, Hong Xiang, Fuliang Zhang
ABSTRACT
The incidence of obesity is increasing worldwide. It was reported that endoplasmic reticulum stress (ERS) could inhibit insulin receptor signaling by activating c-Jun N-terminal kinase (JNK) in the liver. However, the relationship between ERS and insulin receptor signaling in the brain during obesity remains unclear. The aim of the current study was to assess whether ERS alters insulin receptor signaling through the hyper-activation of JNK in the hippocampus and frontal cortex in the brains of obese rats. Obesity was induced using a high fat diet (HFD). The Morris water maze test was then performed to evaluate decreases in cognitive function, and western blot was used to verify whether abnormal insulin receptor signaling was induced by ERS in HFD rats exhibiting cognitive decline. In addition, to determine whether ERS activated JNK and consequently impaired insulin receptor signaling, SH-SY5Y cells were treated with the JNK inhibitor, SP600125, followed by tunicamycin or thapsigargin, and primary rat hippocampal and cortical neurons were transfected with siRNA against IRE1α and JNK. We found that the expression of phosphorylation of PKR-like kinase (PERK), phosphorylation of α subunit of translation initiation factor 2 (eIF2α), and phosphorylation of inositol-requiring kinase-1α (IRE-1α) were increased in the brains of rats with HFD when compared with control rats. The level of serine phosphorylation of insulin receptor substrate-1 (IRS-1) was also increased, while protein kinase B (PKB/Akt) was reduced. ERS was also found to inhibit insulin receptor signaling via the activation of JNK in SH-SY5Y cells, primary rat hippocampal, and cortical neurons. These results indicate that ERS was increased, thereby resulting in impaired insulin receptor signaling in the hippocampus and frontal cortex of obese rats.
Sigma-Aldrich
MISSION® esiRNA, targeting human ERN1
Sigma-Aldrich
MISSION® esiRNA, targeting human MAPK8
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mapk8
Sigma-Aldrich
Anti-IRS1 Antibody, Upstate®, from rabbit