Skip to Content
Merck

Skip To

D9434

Dextrose

97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications

Dextrose

Synonym(s):

D-(+)-Glucose, Dextrose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C6H12O6
CAS Number:
Molecular Weight:
180.16
NACRES:
NA.21
PubChem Substance ID:
UNSPSC Code:
12352201
EC Number:
200-075-1
Beilstein/REAXYS Number:
1724615
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

corn

Quality Segment

grade

anhydrous

agency

USP/NF, meets BP testing specifications, meets EP testing specifications, meets JP testing specifications, meets USP testing specifications

assay

97.5-102.0% anhydrous basis

form

solid

optical activity

[α]/D 52.5 to 53.3 °

quality

meets EP, BP, JP, USP testing specifications

impurities

≤4 ppm heavy metals

color

white

mp

150-152 °C (lit.)

solubility

water: ~470 g/L

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Gene Information

human ... PYGM(5837)

Application

Dextrose has been used:
  • as a component in a basic medium to incubate goat mammary epithelial cells (GMEC) for glucose stimulation
  • in mice to perform glucose-stimulated insulin secretion test (IST) and to quantify serum insulin by enzyme-linked immunosorbent assay (ELISA)
  • to perform glucose tolerance test (GTT) in thermoneutral (TN) and heat-stressed (HS) dairy ewes to study its effects on the feed intake

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G8769G8644RDD016
description

97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications

description

45% in H2O, sterile-filtered, BioXtra, suitable for cell culture

description

100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture

description

≥99.5% (anhydrous), free-flowing, Redi-Dri

biological source

corn

biological source

-

biological source

-

biological source

maize

grade

anhydrous

grade

-

grade

-

grade

-

assay

97.5-102.0% anhydrous basis

assay

-

assay

-

assay

≥99.5% (anhydrous)

Quality Level

200

Quality Level

200

Quality Level

400

Quality Level

100

solubility

water: ~470 g/L

solubility

-

solubility

-

solubility

water: 0.1333 g/mL, clear, colorless

form

solid

form

solution

form

solution

form

powder


Still not finding the right product?

Explore all of our products under Dextrose


Storage Class

11 - Combustible Solids

ppe

Eyeshields, Gloves, type N95 (US)

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. what is the storage temperature for product Dextrose D9434?

    1 answer
    1. As listed in the 'PROPERTIES' section, this product should be stored at room temperature.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review