Merck
  • Role of Organic Coatings in Regulating N2O5 Reactive Uptake to Sea Spray Aerosol.
Role of Organic Coatings in Regulating N2O5 Reactive Uptake to Sea Spray Aerosol.
The journal of physical chemistry. A (2015-11-07)
Olivia S Ryder, Nicole R Campbell, Holly Morris, Sara Forestieri, Matthew J Ruppel, Christopher Cappa, Alexei Tivanski, Kimberly Prather, Timothy H Bertram
ABSTRACT
Previous laboratory measurements and field observations have suggested that the reactive uptake of N2O5 to sea spray aerosol particles is a complex function of particle chemical composition and phase, where surface active organics can suppress the reactive uptake by up to a factor of 60. To date, there are no direct studies of the reactive uptake of N2O5 to nascent sea spray aerosol that permit assessment of the role that organic molecules present in sea spray aerosol (SSA) may play in suppressing or enhancing N2O5 uptake kinetics. In this study, SSA was generated from ambient seawater and artificial seawater matrices using a Marine Aerosol Reference Tank (MART), capable of producing nascent SSA representative of ambient conditions. The reactive uptake coefficient of N2O5 (γ(N2O5)) on nascent SSA was determined using an entrained aerosol flow reactor coupled to a chemical ionization mass spectrometer for measurement of surface area dependent heterogeneous loss rates. Population averaged measurements of γ(N2O5) for SSA generated from salt water sequentially doped with representative organic molecular mimics, or from ambient seawater, do not deviate statistically from that observed for sodium chloride (γ(N2O5)NaCl = 0.01-0.03) for relative humidity (RH) ranging between 50 and 65%. The results are consistent with measurements made under clean marine conditions at the Scripps Institution of Oceanography Pier and those conducted on nascent SSA generated in the marine aerosol reference tank. The results presented here suggest that organic films present on nascent SSA (at RH greater than 50%) likely do not significantly limit N2O5 reactive uptake.
Product Description
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by trichloroacetic acid extraction
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphate sodium salt, ≥99%
SAFC
Cholesterol, Plant-Derived, SyntheChol®