Merck
  • MCT1 promotes the cisplatin-resistance by antagonizing Fas in epithelial ovarian cancer.
MCT1 promotes the cisplatin-resistance by antagonizing Fas in epithelial ovarian cancer.
International journal of clinical and experimental pathology (2015-06-06)
Chunxiao Yan, Fan Yang, Chunxia Zhou, Xuejun Chen, Xuechuan Han, Xueqin Liu, Hongyun Ma, Wei Zheng
ABSTRACT
This study was designed to investigate the role of MCT1 in the development of cisplatin-resistant ovarian cancer and its possible relationship with Fas. We found the expression of MCT1 was obviously increased both in cisplatin-resistant ovarian cancer tissue and A2780/CP cells compared with sensitive ovarian cancer tissue and cell lines A2780. And in A2780 cells treated with Cisplatin, the expression of MCT1 increased in a concentration-dependent manner, MCT1 knockdown attenuates cisplatin-induced cell viability. In A2780 and A2780/CP cells transfected with MCT1 siRNA, the activation of several downstream targets of Fas, including FasL and FAP-1 were largely prevented, whereas the expression of Caspase-3 was increased, accompanying with increased abundance of Fas. Coimmunoprecipitation and immunofluorescence showed that there is interaction between endogenous MCT1 with Fas in vivo and in vitro. In vivo, depletion of MCT1 by shRNA reverses cisplatin-resistance and the expression of Fas. This study showed that down regulation of MCT1 promote the sensibility to Cisplatin in ovarian cancer cell line. And this effect appeared to be mediated via antagonizing the effect of Fas.
Sigma-Aldrich
MISSION® esiRNA, targeting human MCTS1
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mcph1
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human MCAT
Sigma-Aldrich
MISSION® esiRNA, targeting human CMA1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mcat
Sigma-Aldrich
MISSION® esiRNA, targeting human SLC16A1
Sigma-Aldrich
MISSION® esiRNA, targeting human MCPH1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mcts1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Slc16a1