Merck
  • Selection of high-affinity Centyrin FN3 domains from a simple library diversified at a combination of strand and loop positions.
Selection of high-affinity Centyrin FN3 domains from a simple library diversified at a combination of strand and loop positions.
Protein engineering, design & selection : PEDS (2014-05-03)
Michael D Diem, Linus Hyun, Fang Yi, Randi Hippensteel, Elise Kuhar, Cassandra Lowenstein, Edward J Swift, Karyn T O'Neil, Steven A Jacobs
ABSTRACT
Alternative scaffold molecules represent a class of proteins important to the study of protein design and mechanisms of protein-protein interactions, as well as for the development of therapeutic proteins. Here, we describe the generation of a library built upon the framework of a consensus FN3 domain sequence resulting in binding proteins we call Centyrins. This new library employs diversified positions within the C-strand, CD-loop, F-strand and FG-loop of the FN3 domain. CIS display was used to select high-affinity Centyrin variants against three targets; c-MET, murine IL-17A and rat TNFα and scanning mutagenesis studies were used to define the positions of the library most important for target binding. Contributions from both the strand and loop positions were noted, although the pattern was different for each molecule. In addition, an affinity maturation scheme is described that resulted in a significant improvement in the affinity of one selected Centyrin variant. Together, this work provides important data contributing to our understanding of potential FN3 binding interfaces and a new tool for generating high-affinity scaffold molecules.
Sigma-Aldrich
Hepatocyte Growth Factor human, HGF, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Sigma-Aldrich
Hepatocyte Growth Factor human, HGF, recombinant, expressed in Baculovirus infected High-5 cells, suitable for cell culture
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Hepatocyte Growth Factor human, HGF, recombinant, expressed in NSO cells, suitable for cell culture
Sigma-Aldrich
Rat CINC-1 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
HGF human, recombinant, expressed in CHO cells, ≥97% (SDS-PAGE), ≥97% (HPLC)
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
USP
Ampicillin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Human Procalcitonin ELISA Kit
Sigma-Aldrich
Mouse KC / CXCL1 ELISA Kit, for serum, plasma and cell culture supernatant
Supelco
Ampicillin, analytical standard
Sigma-Aldrich
Human GRO-α / CXCL1 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Ampicillin, anhydrous, European Pharmacopoeia (EP) Reference Standard