Merck
  • ERK phosphorylation and miR-181a expression modulate activation of human memory TH17 cells.
ERK phosphorylation and miR-181a expression modulate activation of human memory TH17 cells.
Nature communications (2015-03-17)
Federico Mele, Camilla Basso, Cristina Leoni, Dominik Aschenbrenner, Simone Becattini, Daniela Latorre, Antonio Lanzavecchia, Federica Sallusto, Silvia Monticelli
ABSTRACT
T helper (TH) cell polarization during priming is modulated by a number of signals, but whether polarization to a given phenotype also influences recall responses of memory TH cells is relatively unknown. Here we show that miR-181a is selectively induced in both human and mouse naive T cells differentiating into the TH17, but not TH1 or TH2 subset. In human memory TH17 cells, miR-181a regulates responses to cognate antigens through modulation of ERK phosphorylation. By enhancing the signalling cascade from the T-cell receptor, such molecular network reduces the threshold of TH17 cell activation. Moreover, at a late time point, the same network induces a self-regulatory mechanism dependent on ID3, a negative regulator of transcription factors that control RORC expression, thus modulating TH17 activity. Our results demonstrate that the phenotype acquired by TH cells during priming contributes to their threshold of activation to secondary antigenic stimulations, thus influencing memory responses.
Product Description
Supelco
2-Butanone, analytical standard
Sigma-Aldrich
2-Butanone, FCC, FG
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Butanone, suitable for HPLC, ≥99.7%
Sigma-Aldrich
2-Butanone, ReagentPlus®, ≥99%
Supelco
Methyl Ethyl Ketone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioXtra, ≥98.0%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder