Merck
  • Structural attributes of model protein formulations prepared by rapid freeze-drying cycles in a microscale heating stage.
Structural attributes of model protein formulations prepared by rapid freeze-drying cycles in a microscale heating stage.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-03-13)
Björn-Hendrik Peters, Ferdinand Molnár, Jarkko Ketolainen
ABSTRACT
Downscaled freeze-drying was demonstrated to be a valuable alternative for formulation development and optimization. Although the pore structure is known to exert a major influence on the freeze-drying cycle, little is known about the ones of microscale preparations. This study describes morphology evaluation methods for lysozyme formulations prepared in one microscale processing option and the assessment of fundamental product quality criteria. Scanning electron microscopy (SEM) revealed cooling rate dependent pore size variations at the nucleation site which diminished as the rate increased. Micro-X-ray computed tomography (μ-CT) showed that porosity generally increased in the sample from bottom to top, the pore size fractions shifted toward larger pores in elevated sample levels, and horizontal homogeneity was found throughout each sample with minor deviations in the bottom region. Furthermore, the event of microcollapse could be identified and quantified. Low residual moisture was achieved repeatedly and the procedure did not influence the post freeze-drying bioactivity. This microscale heating stage is a valuable option to reduce overall cycle times and cost, and to prepare freeze-drying formulations with high reproducibility. The mapping tools permit a quick but detailed insight into the structural features resulting from the process environment and processing conditions.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, NMR reference standard
Mannitol, European Pharmacopoeia (EP) Reference Standard
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-(+)-Trehalose dihydrate, ≥99% (HPLC), from starch
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Trehalose dihydrate, from Saccharomyces cerevisiae, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
D-(+)-Trehalose dihydrate, ≥99% (HPLC), from Saccharomyces cerevisiae
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Trehalose dihydrate, ≥98.5% (HPLC), from Saccharomyces cerevisiae
Trehalose dihydrate, European Pharmacopoeia (EP) Reference Standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard