Merck
  • Application of electro-enhanced solid phase microextraction combined with gas chromatography-mass spectrometry for the determination of tricyclic antidepressants in environmental water samples.
Application of electro-enhanced solid phase microextraction combined with gas chromatography-mass spectrometry for the determination of tricyclic antidepressants in environmental water samples.
Journal of chromatography. A (2014-05-28)
Ruyi Xu, Hian Kee Lee
ABSTRACT
A fast and efficient method for the determination of tricyclic antidepressants using electro-enhanced solid phase microextraction (SPME) coupled to gas chromatography-mass spectrometry (GC-MS) has been developed. One advantage of this approach is that the mass transfer of target analytes from the sample solution to an SPME fiber can be accelerated by the electrical field, improving extraction selectivity and efficiency. In the present work, the target analytes extracted to the SPME fiber were thermally desorbed in the GC injection port after extraction. Under the optimized extraction conditions, the developed method exhibited low limits of detection (between 0.079 and 0.296μg/L) and good linearity over the concentration range of between 1 and 500μg/L with coefficients of determination (r(2)) of between 0.993 and 0.999. The relative standard deviations were lower than 9.2% for all analytes. The proposed method was applied to extract tricyclic antidepressants in environmental water samples.
Product Description
USP
Amitriptyline hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Amitriptyline hydrochloride, ≥98% (TLC), powder
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Divinylbenzene, technical grade, 55%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Supelco
Amitriptyline hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Divinylbenzene, technical grade, 80%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Amitriptyline hydrochloride, European Pharmacopoeia (EP) Reference Standard
Clomipramine hydrochloride, for microbiological assay, European Pharmacopoeia (EP) Reference Standard