Merck
  • Anti-Bv8 antibody and metronomic gemcitabine improve pancreatic adenocarcinoma treatment outcome following weekly gemcitabine therapy.
Anti-Bv8 antibody and metronomic gemcitabine improve pancreatic adenocarcinoma treatment outcome following weekly gemcitabine therapy.
Neoplasia (New York, N.Y.) (2014-06-25)
Erez Hasnis, Dror Alishekevitz, Svetlana Gingis-Veltski, Rotem Bril, Ella Fremder, Tali Voloshin, Ziv Raviv, Amir Karban, Yuval Shaked
ABSTRACT
Weekly gemcitabine therapy is the major treatment offered for patients with pancreatic adenocarcinoma cancer; however, relative resistance of tumor cells to chemotherapy, rapid regrowth, and metastasis are the main causes of death within a year. Recently, the daily continuous administration of chemotherapy in low doses--called metronomic chemotherapy (MC)--has been shown to inhibit primary tumor growth and delay metastases in several tumor types; however, its use as a single therapy is still in question due to its moderate therapeutic benefit. Here, we show that the combination of weekly gemcitabine with MC of the same drug delays tumor regrowth and inhibits metastasis in mice implanted orthotopically with pancreatic tumors. We further demonstrate that weekly gemcitabine, but not continuous MC gemcitabine or the combination of the two drug regimens, promotes rebound myeloid-derived suppressor cell (MDSC) mobilization and increases angiogenesis in this tumor model. Furthermore, Bv8 is highly expressed in MDSCs colonizing pancreatic tumors in mice treated with weekly gemcitabine compared to MC gemcitabine or the combination of the two regimens. Blocking Bv8 with antibodies in weekly gemcitabine-treated mice results in a significant reduction in tumor regrowth, angiogenesis, and metastasis. Overall, our results suggest that pro-tumorigenic effects induced by weekly gemcitabine are mediated in part by MDSCs expressing Bv8. Therefore, both Bv8 inhibition and MC can be used as legitimate 'add-on' treatments for preventing post-chemotherapy pancreatic cancer recurrence, progression, and metastasis following weekly gemcitabine therapy.
Product Description
USP
Gemcitabine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Gemcitabine hydrochloride, ≥98% (HPLC)
Gemcitabine hydrochloride, European Pharmacopoeia (EP) Reference Standard