Merck
  • Pituitary adenylate cyclase-activating polypeptide (PACAP) signalling enhances osteogenesis in UMR-106 cell line.
Pituitary adenylate cyclase-activating polypeptide (PACAP) signalling enhances osteogenesis in UMR-106 cell line.
Journal of molecular neuroscience : MN (2014-08-13)
Tamás Juhász, Csaba Matta, Éva Katona, Csilla Somogyi, Roland Takács, Tibor Hajdú, Solveig Lind Helgadottir, János Fodor, László Csernoch, Gábor Tóth, Éva Bakó, Dóra Reglődi, Andrea Tamás, Róza Zákány
ABSTRACT
Presence of the pituitary adenylate cyclase-activating polypeptide (PACAP) signalling has been proved in various peripheral tissues. PACAP can activate protein kinase A (PKA) signalling via binding to pituitary adenylate cyclase-activating polypeptide type I receptor (PAC1), vasoactive intestinal polypeptide receptor (VPAC) 1 or VPAC2 receptor. Since little is known about the role of this regulatory mechanism in bone formation, we aimed to investigate the effect of PACAP on osteogenesis of UMR-106 cells. PACAP 1-38 as an agonist and PACAP 6-38 as an antagonist of PAC1 were added to the culture medium. Surprisingly, both substances enhanced protein expressions of collagen type I, osterix and alkaline phosphatase, along with higher cell proliferation rate and an augmented mineralisation. Although expression of PKA was elevated, no alterations were detected in the expression, phosphorylation and nuclear presence of CREB, but increased nuclear appearance of Runx2, the key transcription factor of osteoblast differentiation, was shown. Both PACAPs increased the expressions of bone morphogenetic proteins (BMPs) 2, 4, 6, 7 and Smad1 proteins, as well as that of Sonic hedgehog, PATCH1 and Gli1. Data of our experiments indicate that activation of PACAP pathway enhances bone formation of UMR-106 cells and PKA, BMP and Hedgehog signalling pathways became activated. We also found that PACAP 6-38 did not act as an antagonist of PACAP signalling in UMR-106 cells.
Sigma-Aldrich
Anti-SMAD1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, tablet
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Anti-BMP4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution