Merck
  • Single gene insertion drives bioalcohol production by a thermophilic archaeon.
Single gene insertion drives bioalcohol production by a thermophilic archaeon.
Proceedings of the National Academy of Sciences of the United States of America (2014-11-05)
Mirko Basen, Gerrit J Schut, Diep M Nguyen, Gina L Lipscomb, Robert A Benn, Cameron J Prybol, Brian J Vaccaro, Farris L Poole, Robert M Kelly, Michael W W Adams
ABSTRACT
Bioethanol production is achieved by only two metabolic pathways and only at moderate temperatures. Herein a fundamentally different synthetic pathway for bioalcohol production at 70 °C was constructed by insertion of the gene for bacterial alcohol dehydrogenase (AdhA) into the archaeon Pyrococcus furiosus. The engineered strain converted glucose to ethanol via acetate and acetaldehyde, catalyzed by the host-encoded aldehyde ferredoxin oxidoreductase (AOR) and heterologously expressed AdhA, in an energy-conserving, redox-balanced pathway. Furthermore, the AOR/AdhA pathway also converted exogenously added aliphatic and aromatic carboxylic acids to the corresponding alcohol using glucose, pyruvate, and/or hydrogen as the source of reductant. By heterologous coexpression of a membrane-bound carbon monoxide dehydrogenase, CO was used as a reductant for converting carboxylic acids to alcohols. Redirecting the fermentative metabolism of P. furiosus through strategic insertion of foreign genes creates unprecedented opportunities for thermophilic bioalcohol production. Moreover, the AOR/AdhA pathway is a potentially game-changing strategy for syngas fermentation, especially in combination with carbon chain elongation pathways.
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
Uracil, ≥99.0%
Sigma-Aldrich
Sodium acetate-13C2, 99 atom % 13C
Sigma-Aldrich
Maltose solution, for molecular biology, BioReagent, ~20% in H2O
Supelco
Uracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium acetate-13C2, endotoxin tested, 99 atom % 13C
Fluorouracil impurity C, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.