Merck
  • Acute and chronic effects of IL-22 on acetaminophen-induced liver injury.
Acute and chronic effects of IL-22 on acetaminophen-induced liver injury.
Journal of immunology (Baltimore, Md. : 1950) (2014-07-27)
Dechun Feng, Yan Wang, Hua Wang, Honglei Weng, Xiaoni Kong, Brittany V Martin-Murphy, Yongmei Li, Ogyi Park, Steven Dooley, Cynthia Ju, Bin Gao
ABSTRACT
Acetaminophen (APAP)-induced liver injury (AILI) accounts for half of the acute liver failure cases in the United States. A better understanding of the underlying mechanisms of AILI is necessary for the development of novel antidotes. We found that pretreatment with IL-22 protected mice from APAP-mediated hepatotoxicity. The protection was dependent on STAT3, as IL-22 failed to reduce APAP hepatotoxicity in liver-specific STAT3 knockout mice. In contrast to the acute exposure to IL-22, the endogenous chronic overexpression of IL-22 in IL-22 transgenic (TG) mice or IL-22 adenovirus treatment for 6 wk resulted in a markedly increased susceptibility to AILI. Furthermore, the hepatic expression levels of cytochrome 2E1 (Cyp2E1) and Cyp1A2 were much higher in IL-22TG mice. Ablation of Cyp2E1 but not hepatic STAT3 abolished AILI and protein-adduct formation in IL-22TG mice. Finally, hepatic expression of HNF-1α, a transcriptional factor that is known to control Cyp2E1 expression, was elevated in IL-22TG mice compared with wild-type mice. Upregulation of hepatic Cyp2E1 was only observed in mice with constitutive overexpression of IL-22 but not with short-term treatment with one dose of IL-22 or multiple doses of IL-22 for 2 wk. In conclusion, short-term acute IL-22 exposure protects mice against AILI through STAT3 activation; however, chronic constitutive overexpression of IL-22 exacerbates AILI by increasing Cyp2E1 and toxic reactive APAP metabolite production. These findings may not only enhance our understanding of the effects of chronic inflammation on AILI in patients with liver disease, but are also helpful to identify novel therapeutic targets for the treatment of AILI.
Product Description
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
IL-22 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard