Merck
  • Identification of odorants in frankincense (Boswellia sacra Flueck.) by aroma extract dilution analysis and two-dimensional gas chromatography-mass spectrometry/olfactometry.
Identification of odorants in frankincense (Boswellia sacra Flueck.) by aroma extract dilution analysis and two-dimensional gas chromatography-mass spectrometry/olfactometry.
Phytochemistry (2014-12-04)
Johannes Niebler, Andrea Buettner
ABSTRACT
Frankincense has been known, traded and used throughout the ages for its exceptional aroma properties, and is still commonly used in both secular and religious settings to convey a pleasant odor. Surprisingly, the odoriferous principle(s) underlying its unique odor profile have never been published. In this study, resin samples of Boswellia sacra Flueck. from both Somalia and Oman were investigated by aroma extract dilution analysis. In a comprehensive, odor-activity guided approach both chemo-analytical and human-sensory parameters were used to identify odor active constituents of the volatile fraction of B. sacra. Among the key odorants found were α-pinene, β-myrcene, linalool, p-cresol and two unidentified sesquiterpenoids. Overall, a total of 23 odorants were detected and analyzed by gas chromatography-olfactometry and heart-cut two-dimensional gas chromatography-mass spectrometry/olfactometry. The majority of the identified odorant compounds were oxygenated monoterpenes, along with some relevant mono- and sesquiterpenes and only one diterpenoid substance. Several of these compounds were reported here for the first time as odorous constituents in B. sacra. Identifying bioactive compounds might support a better understanding with regard to the potential benefits of frankincense, for example in aromatherapy or ecclesial settings.
Product Description
Supelco
1,8-Cineole, primary reference standard
Supelco
Hexane, analytical standard
Supelco
Tetratriacontane, analytical standard
Sigma-Aldrich
Ethyl 2-methylbutyrate, natural, mixture of enantiomers, ≥98%, FCC, FG
Sigma-Aldrich
Ethyl 2-methylbutyrate, 99%
Sigma-Aldrich
2-Methylanisole, ≥99%, FG
Sigma-Aldrich
4-Methylanisole, 99%
Sigma-Aldrich
Ethyl 2-methylbutyrate, ≥98%, FCC, FG
Sigma-Aldrich
Ethyl isovalerate, ≥98%, FCC, FG
Sigma-Aldrich
Pentane, anhydrous, ≥99%
Sigma-Aldrich
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
Supelco
m-Cresol, analytical standard
Sigma-Aldrich
Ethyl isovalerate, natural, ≥98%, FCC, FG
Sigma-Aldrich
2-Methylanisole, 99%
Supelco
Pentane, analytical standard
Sigma-Aldrich
Ethyl isovalerate, 98%
Sigma-Aldrich
o-Cymene, 98%
Sigma-Aldrich
m-Cresol, ≥98%, FG
Supelco
(−)-Linalool, analytical standard
Sigma-Aldrich
Pentane, ≥99% (GC)
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
m-Cresol, 99%
Sigma-Aldrich
Pentane, reagent grade, 98%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Pentane, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Pentane, spectrophotometric grade, ≥99%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%