Skip to Content
Merck
All Photos(2)

Key Documents

139386

Sigma-Aldrich

Hexane

≥99%, suitable for GC/MS, suitable for EPA 1613, ReagentPlus®

Synonym(s):

n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.06
grade:
reagent
technique(s):
GC/MS: suitable
bp:
69 °C (lit.)
vapor pressure:
256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

Product Name

Hexane, ReagentPlus®, ≥99%

grade

reagent

Quality Level

Agency

suitable for EPA 1613

vapor density

~3 (vs air)

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

453 °F

expl. lim.

7.7 %

technique(s)

GC/MS: suitable

refractive index

n20/D 1.375 (lit.)

pH

7

bp

69 °C (lit.)

mp

−95 °C (lit.)

solubility

water: insoluble

density

0.659 g/mL at 25 °C (lit.)

SMILES string

CCCCCC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexane is an alkane and exists in 5 structural isomeric forms, i.e. hexane, isohexane, 3-methylpentane, neohexane and 2,3-dimethylbutane. It can be obtained from petroleum by distillation. It can also be produced by electrolyzing the potassium butyrate solution. It is widely employed organic solvent for the dissolution and isolation of fats and oils.

Application

Hexane may be used in the following studies:
  • Preparation of graft-copolymer chains of poly-(dimethylsiloxane)-g-poly(methyl methacrylate).
  • GC/MS (Gas Chromatography/Mass Spectrometry) characterization of essential oils isolated from various origins.
  • As solvent for the preparation of samples and standards during chromatographic studies.

Hexane may be used as a solvent in the detection of rubrene, using normal phase high performance liquid chromatography. It may also be used as an extraction solvent to concentrate the analytes like synthetic cannabinoids of the aminoalkylindole type and methanandamide in serum, followed by separation using liquid chromatography coupled with tandem-mass spectrometry.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Development and validation of a liquid chromatography?tandem mass spectrometry method for the quantitation of synthetic cannabinoids of the aminoalkylindole type and methanandamide in serum and its application to forensic samples
Dresen S, et al.
Journal of Mass Spectrometry : Jms, 46(2), 163-171 (2011)
Occurrence, mechanisms and analytical applications of electrogenerated chemiluminescence. A review
Knight.WA and Greenway.MG et al.
Analyst, 119(5), 879- 890 (1994)
G Di Nicola et al.
Journal of chromatography. A, 1190(1-2), 120-126 (2008-03-18)
Biodiesel (a mixture of fatty acid esters) is normally analyzed using gas chromatography/flame ionization detection, as specified by the ASTM D6584 and EN14105 standards. This paper proposes a binary gradient method for analyzing biodiesel mixtures using non-aqueous reverse phase HPLC
Comparative evaluation of 11 essential oils of different origin as functional antioxidants, antiradicals and antimicrobials in foods.
Sacchetti G, et al.
Food Chemistry, 91(4), 621-632 (2005)
Eagleson M.
Concise Encyclopedia Chemistry, 492-492 (1994)

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service