Merck
  • Silver nanoparticles supported on passivated silica: preparation and catalytic performance in alkyne semi-hydrogenation.
Silver nanoparticles supported on passivated silica: preparation and catalytic performance in alkyne semi-hydrogenation.
Dalton transactions (Cambridge, England : 2003) (2014-09-03)
Emma Oakton, Gianvito Vilé, Daniel S Levine, Eva Zocher, David Baudouin, Javier Pérez-Ramírez, Christophe Copéret
ABSTRACT
Herein, we report the preparation of small and narrowly distributed (2.1 ± 0.5 nm) Ag nanoparticles supported on passivated silica, where the surface OH groups are replaced by OSiMe3 functionalities. This synthetic method involves the grafting of silver(I) bis(trimethylsilyl)amide ([AgN(SiMe3)2]4) on silica partially dehydroxylated at 700 °C, followed by a thermal treatment of the grafted complex under H2. The catalytic performance of this material was investigated in the semi-hydrogenation of propyne and 1-hexyne and compared with that of 2.0 ± 0.3 nm Ag nanoparticles supported on silica. Whilst surface passivation slightly decreases the activity in both reactions (by a factor 2-3), probably as a result of the decreased alkyne adsorption properties or the presence of less accessible active sites on the passivated support, the AgNP@SiO2 catalysts demonstrate a remarkable selectivity for the production of alkenes.
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
1-Hexyne, 97%
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Supelco
Benzene, analytical standard
Supelco
Toluene, analytical standard
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Rhein
Sigma-Aldrich
Rhein, technical grade