Skip to Content
Merck
All Photos(1)

Documents

676756

Sigma-Aldrich

Toluene

ACS reagent, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

997 °F

expl. lim.

7 %

technique(s)

toxicology assay: suitable

impurities

H2SO4, passes test (darkened)
≤0.003% S compounds
≤0.03% Water

evapn. residue

≤0.001%

refractive index

n/D 1.496 (lit.)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

solubility

acetone: miscible(lit.)
alcohol: miscible(lit.)
carbon disulfide: miscible(lit.)
chloroform: miscible(lit.)
glacial acetic acid: miscible(lit.)

density

0.865 g/mL at 25 °C (lit.)

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Toluene, also known as methylbenzene, is a flammable liquid with a pungent odor. It is a constituent of crude oil. Toluene is widely used as a precursor for synthesizing benzene, a gasoline additive, and as a solvent in the paint industry. Its toxicological effects have been investigated. Alkylation of toluene to form p-xylene with high selectivity using modified ZSM (Zeolite Socony Mobil)-5-class zeolite catalysts has been reported. Electrical properties of graphene can be altered by doping it with toluene, which serves as an electron donor. Temperature programmed hydrogenation (TPH) of toluene in the gas phase in the presence of supported metal catalysts has been studied.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F - closed cup

Flash Point(C)

4.4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Temperature programmed hydrogenation of toluene.
Slioor RI, et al.
Catalysis Letters, 121(1-2), 24-32 (2008)
Toluene.
von Burg R.
Journal of Applied Toxicology, 13(6), 441-446 (1993)
Selective alkylation of toluene with methanol to produce para-xylene.
Kaeding WW, et al.
J. Catal., 67(1), 159-174 (1981)
Electrochemical doping of graphene with toluene.
Kaverzin AA, et al.
Carbon, 49(12), 3829-3834 (2011)
Liang Guo et al.
Journal of chromatography. A, 1286, 9-15 (2013-03-19)
For the first time, a novel one-step sample preparation method that combines microwave assisted extraction and solvent bar microextraction (MAE-SBME) with analysis by gas chromatography-mass spectrometry (GC-MS), was developed for the fast and efficient determination of polycyclic aromatic hydrocarbons (PAHs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service