Merck
  • Assessment of the immunogenicity of mechanically induced interferon aggregates in a transgenic mouse model.
Assessment of the immunogenicity of mechanically induced interferon aggregates in a transgenic mouse model.
Journal of pharmaceutical sciences (2014-12-03)
Paul Human, Helen Ilsley, Cynthia Roberson, Eric Grovender, Bill Van Antwerp, Eric Fogt, Peter Zilla
ABSTRACT
Pump delivery of human interferon alpha-2B (IFNα2b) has the potential for inducing immunogenic drug aggregates. We therefore evaluated the immunogenicity of mechanically induced IFNα2b aggregates to assess this risk. Transgenic human-IFNα2b (TG) and wild-type (WT) FVB/N mice (n = 8 and n = 9/group, respectively) were administered mechanically agitated drug [45 Hz for 6 h (LLA) or 24 h (HLA)], chemically modified drug [low pH (pH 4.0) or metal oxidized (OXD)] or unstressed drug (native). Mice received IFNα2b (50 μg; 100 μg/mL; s.c.) formulations on days 0, 7, 14, and 21. Drug-binding and neutralizing antibody titers were determined after 28 d. Aggregate concentrations were highest in OXD and HLA formulations but OXD had more dimers/trimers. Geometric mean titers were 1:131, 1:728, 1:1573, 1:871, and 1:10,240 for WT mice (n = 9) and 1:207, 1:587, 1:1810, 1:571, and 1:2,153 for TG mice (n = 8) for native, LLA, HLA, pH4, and OXD groups, respectively. Mechanical agitation of IFNα2b induced equivalent titers of immunoglobulin to that of metal oxidation, both capable of binding to or neutralizing the drug in WT and TG mice. Thus, by limiting metal contamination and by inclusion of a stabilizing agent to mitigate drug aggregation, the risk of anti-drug immunoglobulin may be reduced in a pump delivery scenario.
Product Description
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium phosphate, 96%
Supelco
Protein Standard, analytical standard, 2 mg/vial BSA
Sigma-Aldrich
Guanidine hydrochloride, for molecular biology, ≥99%